    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
          • Order Status
          • 快速訂購
          • 登入
            登入
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • GLP-1 Antibodies
          Invitrogen
          GLP-1 Monoclonal Antibody (10)
          2 Published Figures
          2 References
          View all (9) GLP-1 antibodies
          Tech Support
          Cite GLP-1 Monoclonal Antibody (10)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          • Published Figures (2)
          GLP-1 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          GLP-1 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          FIGURE: 1 / 5
          GLP-1 Antibody (ABS 033-10-02) in ELISA
          Sandwich ELISA of Glucagon-Like Peptide 1 using ABS 046-03 as the capture antibody and ABS 033-10B as the biotinylated detection antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GLP-1 Antibody in ELISA (ELISA)
          GLP-1 Antibody in ELISA (ELISA)
          GLP-1 Antibody in ELISA (ELISA)
          GLP-1 Antibody in Western Blot (WB)
          GLP-1 Antibody in ELISA (ELISA)
          GLP-1 Monoclonal Antibody (10)
          Product Details
          ABS 033-10-02
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          -
          View 1 publication 1 publication
          ELISA (ELISA)
          1:20,000
          View 1 publication 1 publication
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          10
          Immunogen
          Synthetic glucagon-like peptide-1 (7-17) adsorbed into aluminum hydroxide gel.
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1.02 mg/mL
          Amount
          408 µg
          Purification
          Protein A
          Storage buffer
          PBS, pH 7.4
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_1074637
          Product Specific Information
          ABS 033-10-02 detects the amidated N-terminus of GLP-1(7-37) and GLP-1(7-36) in human samples.
          ABS 033-10-02 has been successfully used in ELISA procedures. ABS 033-10-02 binds the free N-terminus of GLP-1(7-37) and GLP-1(7-36)amide and shows <0.2% cross-reactivity with GLP-1(9-36)amide, glucagon and human GIP. Cross-reacts 0.25% with GLP-1(1-37), approximately 6% with human GLP-2 and approximately 50% with exendin-4.
          NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
          Target Information
          GCG is also known as GLP1, or Glucagon. Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis.It is mapped to 2q36-2q37. GLP1, also known as 7-37 for the codons of the preproglucagon molecule which encode it, renders pancreatic beta-cells 'glucose-competent' and may be useful in the treatment of noninsulin-dependent diabetes mellitus. Also, GLP1 is a potent insulin secretagogue.It plays a major role in the enteroinsular axis, accounting, for example, for the finding that plasma insulin levels accompanying oral intake of glucose are greater than those observed when glucose is given intravenously.
          Bioinformatics
          Protein Aliases: glicentin-related polypeptide; GLP1/2; Glucagon like peptide 1; glucagon-like peptide 1; glucagon-like peptide 2; MGC138331; OXY; preproglucagon; Pro-glucagon
          View more View less
          Gene Aliases: GCG; GLP-1; GLP1; GLP2; GRPP
          View more View less
          UniProt ID: (Human) P01275
          View more View less
          Entrez Gene ID: (Human) 2641
          View more View less
          Function(s)
          receptor binding hormone activity protein binding gastric inhibitory polypeptide receptor binding glucagon receptor binding identical protein binding receptor agonist activity
          Process(es)
          gluconeogenesis G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway feeding behavior protein kinase A signaling response to activity positive regulation of insulin secretion involved in cellular response to glucose stimulus glucose homeostasis negative regulation of apoptotic process positive regulation of gluconeogenesis regulation of insulin secretion positive regulation of ERK1 and ERK2 cascade cellular response to glucagon stimulus positive regulation of calcium ion import negative regulation of execution phase of apoptosis
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline