  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • GLP-1 Antibodies
          Invitrogen
          GLP-1 Monoclonal Antibody (10), Biotin
          1 Published Figure
          1 Reference
          View all (11) GLP-1 antibodies
          Tech Support
          Cite GLP-1 Monoclonal Antibody (10), Biotin
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          • Published Figures (1)
          GLP-1 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          GLP-1 Antibody in ELISA (ELISA)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          GLP-1 Antibody (ABS 033-10B-005) in ELISA
          Sandwich ELISA of Glucagon-Like Peptide 1 using ABS 046-03 as the capture antibody and ABS 033-10B as the biotinylated detection antibody. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GLP-1 Antibody in ELISA (ELISA)
          GLP-1 Antibody in ELISA (ELISA)
          GLP-1 Antibody in Western Blot (WB)
          GLP-1 Monoclonal Antibody (10), Biotin
          Product Details
          ABS 033-10B-005
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          -
          View 1 publication 1 publication
          ELISA (ELISA)
          1:40,000
          -
          Product Specifications
          Species Reactivity
          Human
          Host/Isotype
          Mouse / IgG2a, kappa
          Class
          Monoclonal
          Type
          Antibody
          Clone
          10
          Immunogen
          Synthetic GLP-1(7-17).
          3D Epitope / Immunogen
          Conjugate
          Biotin Biotin Biotin
          • Unconjugated
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1.05 mg/mL
          Amount
          157.5 µg
          Purification
          purified
          Storage buffer
          PBS, pH 7.4
          Contains
          15mM sodium azide
          Storage conditions
          4°C, store in dark, DO NOT FREEZE!
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          RRID
          AB_962097
          Product Specific Information
          ABS 033-10B-005 detects GLP1 from human samples. ABS 033-10B-005 is specific towards the free N-terminus of GLP-1(7-37) and GLP-1(7-36)amide.
          ABS 033-10B-005 has been successfully used in ELISA procedures.
          NOTE: Concentration is lot-dependent and can vary from 0.85-1.15 mg/mL
          Target Information
          GCG is also known as GLP1, or Glucagon. Glucagon is a 29-amino acid pancreatic hormone that counteracts the glucose-lowering action of insulin by stimulating glycogenolysis and gluconeogenesis.It is mapped to 2q36-2q37. GLP1, also known as 7-37 for the codons of the preproglucagon molecule which encode it, renders pancreatic beta-cells 'glucose-competent' and may be useful in the treatment of noninsulin-dependent diabetes mellitus. Also, GLP1 is a potent insulin secretagogue.It plays a major role in the enteroinsular axis, accounting, for example, for the finding that plasma insulin levels accompanying oral intake of glucose are greater than those observed when glucose is given intravenously.
          Bioinformatics
          Protein Aliases: glicentin-related polypeptide; GLP1/2; Glucagon like peptide 1; glucagon-like peptide 1; glucagon-like peptide 2; MGC138331; OXY; preproglucagon; Pro-glucagon
          View more View less
          Gene Aliases: GCG; GLP-1; GLP1; GLP2; GRPP
          View more View less
          UniProt ID: (Human) P01275
          View more View less
          Entrez Gene ID: (Human) 2641
          View more View less
          Function(s)
          receptor binding hormone activity protein binding gastric inhibitory polypeptide receptor binding glucagon receptor binding identical protein binding receptor agonist activity
          Process(es)
          gluconeogenesis G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway feeding behavior protein kinase A signaling response to activity positive regulation of insulin secretion involved in cellular response to glucose stimulus glucose homeostasis negative regulation of apoptotic process positive regulation of gluconeogenesis regulation of insulin secretion positive regulation of ERK1 and ERK2 cascade cellular response to glucagon stimulus positive regulation of calcium ion import negative regulation of execution phase of apoptosis
          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline