    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • ORAI3 Antibodies
          Alomone Labs, LTD
          Orai3 Polyclonal Antibody
          View all (12) ORAI3 antibodies
          Tech Support
          Cite Orai3 Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          Orai3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Orai3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          Orai3 Antibody (ACC-065-50UL) in WB
          Western blot analysis of rat (lanes 1 and 3) and mouse (lanes 2 and 4) heart membranes: - 1,2. Anti-Orai3 Antibody (#ACC-065), (1:800).3,4. Anti-Orai3 Antibody , preincubated with Orai3 Blocking Peptide (#BLP-CC065). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Orai3 Antibody in Western Blot (WB)
          Orai3 Polyclonal Antibody
          Product Details
          ACC-065-50UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:800
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)REFVHRGYLDLMGAS, corresponding to amino acid residues 28-42 of rat Orai3, Intracellular, N-terminus
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          40 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          ORAI3 is a protein coding gene that is involved in calcium signaling and regulation of calcium homeostasis. It is a member of the ORAI family of proteins that form calcium release-activated calcium (CRAC) channels in the plasma membrane of cells. These channels are activated by depletion of calcium from the endoplasmic reticulum, leading to an influx of calcium ions into the cytoplasm. ORAI3 has been implicated in a variety of physiological processes, including T-cell activation, muscle contraction, and neurotransmitter release. Dysregulation of ORAI3 expression or function has been associated with various diseases, including immune disorders, cancer, and cardiovascular disease
          Bioinformatics
          Protein Aliases: calcium release-activated calcium channel 3; calcium release-activated calcium modulator 3 protein; Protein orai-3; Transmembrane protein 142C; unnamed protein product
          View more View less
          Gene Aliases: 9930124N15; BC061259; ORAI3; RGD1306538; TMEM142C
          View more View less
          UniProt ID: (Human) Q9BRQ5, (Mouse) Q6P8G8, (Rat) Q6AXR8
          View more View less
          Entrez Gene ID: (Human) 93129, (Mouse) 269999, (Rat) 309000
          View more View less
          Function(s)
          calcium channel activity protein binding store-operated calcium channel activity molecular_function ion channel transporter
          Process(es)
          store-operated calcium entry ion transport calcium ion transport ion transmembrane transport calcium ion transmembrane transport biological_process
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline