    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • TRPML2 Antibodies
          Alomone Labs, LTD
          TRPML2 (Mucolipin-2) Polyclonal Antibody
          View all (10) TRPML2 antibodies
          Tech Support
          Cite TRPML2 (Mucolipin-2) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          TRPML2 (Mucolipin-2) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          TRPML2 (Mucolipin-2) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          TRPML2 (Mucolipin-2) Antibody (ACC-082-25UL) in ICC/IF
          Expression of TRPML2 in mouse muscle myoblast (C2C12) cell line - Immunocytochemical staining of TRPML2 in C2C12 cells. A. Cells were stained with Anti-TRPML2 (Mucolipin-2) Antibody (#ACC-082), (red), (1:50) followed by goat Anti-rabbit-AlexaFluor-594 secondary Antibody . B. DAPI is used as the counter staining (blue). C. Merged images of A and B. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          TRPML2 (Mucolipin-2) Antibody in Immunocytochemistry (ICC/IF)
          TRPML2 (Mucolipin-2) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          TRPML2 (Mucolipin-2) Antibody in Western Blot (WB)
          TRPML2 (Mucolipin-2) Polyclonal Antibody
          Product Details
          ACC-082-25UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          Assay-dependent
          -
          Immunocytochemistry (ICC/IF)
          1:50
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)RLDFYRLVQVDISFALK, corresponding to amino acid residues 212-228 of mouse TRPML2, Intracellular, N-terminus
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          20 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          Mucolipins constitute a family of cation channel proteins with homologs in mouse, Drosophila, and C. elegans. Mutations in the human MCOLN1 gene cause mucolipodosis IV.
          Bioinformatics
          Protein Aliases: FLJ36691; MCLN2; mucolipidin 2; mucolipin 2; Mucolipin-2; Transient receptor potential channel mucolipin 2; TRPML2; unnamed protein product
          View more View less
          Gene Aliases: 3300002C04Rik; AI549968; C86638; MCOLN2; TRP-ML2; TRPML2
          View more View less
          UniProt ID: (Human) Q8IZK6, (Mouse) Q8K595
          View more View less
          Entrez Gene ID: (Human) 255231, (Mouse) 68279, (Rat) 292168
          View more View less
          Function(s)
          cation channel activity calcium channel activity iron ion transmembrane transporter activity identical protein binding NAADP-sensitive calcium-release channel activity ion channel
          Process(es)
          adaptive immune response immune system process ion transport calcium ion transport protein transport calcium-mediated signaling positive regulation of chemokine production ion transmembrane transport iron ion transmembrane transport innate immune response calcium ion transmembrane transport positive regulation of monocyte chemotactic protein-1 production positive regulation of macrophage inflammatory protein 1 alpha production positive regulation of chemokine (C-C motif) ligand 5 production macrophage migration neutrophil migration regulation of chemokine (C-X-C motif) ligand 2 production positive regulation of chemokine (C-X-C motif) ligand 2 production transport
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline