Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LLEDLVKKK IHNANPELTD GQIQAMLRRW FVEVEGTVKA YVWDNNKDLA EWLEKQLTEE DGVHSVIEEN IKCISRDYVL KQIRSLVQAN PEVAMDSIIH MTQHISPTQR AEVIRILSTM DSPST (2260-2383 aa encoded by BC137287 )
Acetyl-CoA carboxylase (ACC) is a complex multifunctional enzyme system. ACC is a biotin-containing enzyme which catalyzes the carboxylation of acetyl-CoA to malonyl-CoA, the rate-limiting step in fatty acid synthesis. There are two ACC forms, alpha and beta, encoded by two different genes. ACC-alpha is highly enriched in lipogenic tissues. The enzyme is under long term control at the transcriptional and translational levels and under short term regulation by the phosphorylation/dephosphorylation of targeted serine residues and by allosteric transformation by citrate or palmitoyl-CoA. Multiple alternatively spliced transcript variants divergent in the 5' sequence and encoding distinct isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Acaca; ACC-alpha; ACC1; acetyl-CoA carboxylase (EC 6.4.1.2); acetyl-CoA carboxylase 265; acetyl-coenzyme A carboxylase (EC 6.4.1.2); acetyl-Coenzyme A carboxylase alpha; EC 6.3.4.14; EC 6.4.1.2; unnamed protein product
Gene Aliases: A530025K05Rik; ACAC; Acac1; ACACAD; ACACalpha; ACC; ACC1; ACCA; ACCalpha; Gm738; hACC1
UniProt ID: (Human) Q13085, (Mouse) Q5SWU9, (Rat) P11497
Entrez Gene ID: (Human) 31, (Mouse) 107476, (Rat) 60581
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support