Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 5
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Recommended positive controls: 293T, A431, Jurkat, Raji.
Predicted reactivity: Mouse (95%), Rat (95%), Pig (95%), Bovine (96%).
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
An initial reaction in fatty acid metabolism in eukaryotic cells is activation of fatty acids catalyzed by acyl-CoA synthetase. FACL3 (fatty acid CoA ligase, long-chain 3) is identified as member of the acyl-CoA synthetase (ACS) family by PCR of rat brain cDNAs using primers based on the conserved region of the ACS protein. The 720-amino acid rat protein preferentially utilizes myristate, laurate, arachidonate, and eicosapentaenoate, and is expressed primarily in brain. The predicted 720-amino acid FACL3 human protein is 92% identical to that of rat.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arachidonate--CoA ligase; arachidonate--CoA ligase Acsl3; brain acyl-CoA synthetase II; fatty acid Coenzyme A ligase, long chain 3; fatty-acid-Coenzyme A ligase, long-chain 3; lignoceroyl-CoA synthase; long-chain acyl-CoA synthetase 3; long-chain-fatty-acid--CoA ligase 3; medium-chain acyl-CoA ligase Acsl3; predicted protein of HQ2194; unnamed protein product
Gene Aliases: 2610510B12Rik; ACS3; FACL3; LACS 3; LACS3; PRO2194
UniProt ID: (Human) O95573, (Mouse) Q9CZW4, (Rat) Q63151
Entrez Gene ID: (Human) 2181, (Mouse) 74205, (Rat) 114024
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support