Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • ADAMTS5 Antibodies

          Invitrogen

          ADAMTS5 Polyclonal Antibody

          3 Published Figures
          5 References
          View all (10) ADAMTS5 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ADAMTS5 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Published Figures (3)
          ADAMTS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ADAMTS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ADAMTS5 Antibody (PA1-1751A) in IHC (P)

          GeroScience 2024 - Mice with growth hormone receptor deficiency (GHR -/- ) (24 months) are protected from developing aging-associated OA. ( A ) Safranin-O and Fast Green-stained images of knee joints from WT and GHR -/- mice. Low-magnification (4x) of the whole joint is on the left, and high-magnification (20x) of each compartment (lateral femur, medial femur, lateral tibia, medial tibia) is on the right. Scale bar (green rec... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ADAMTS5 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ADAMTS5 Antibody in Immunohistochemistry (IHC)
          ADAMTS5 Antibody in Immunohistochemistry (IHC)
          ADAMTS5 Polyclonal Antibody

          Product Details

          PA1-1751A

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1 µg/mL
          -

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Bovine, Human, Pig

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide corresponding to residues C K(636) N G Y Q S D A K G V K T F(649) of human ADAMTS5.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1070008

          Product Specific Information

          PA1-1751A detects ADAMTS5 from porcine, bovine, and human samples. PA1-1751A is specific for ADAMTS5 and shows no cross-reactivity with other ADAMTS family members.

          PA1-1751A has been successfully used in Western blot procedures. By Western blot, this antibody detects a major band at ~68 kDa protein representing ADAMTS5 from porcine cartilage extract. On human recombinant ADAMTS5 this antibody detects a major band at ~85 kDa. Depending on the extent of C-terminal processing, several smaller bands may also be seen using this antibody.

          PA1-1751A immunogen is a synthetic peptide corresponding to residues C K(636) N G Y Q S D A K G V K T F(649) of human ADAMTS5. This polyclonal antibody has been referred to as: JSCKNG.

          Target Information

          ADAMTS5, also known as Implantin or Aggrecanase 2, is a member of the larger family of ADAMs (A Disintegrin And Metalloproteinase) metalloproteinases containing thrombospondin (TS) repeats. ADAMTS5 (A Disintegrin And Metalloproteinase with Thrombospondin-5 motif) was first described as ADAMTS5, a protein elevated in mice during the peri-implantion period. At the same time, another group identified Aggrecanase 11, a protein elevated in arthritic synovium. The name was later changed to ADAMTS5. ADAMTS5 is expressed in human and mouse. It has been found in heart, lung, cervix, uterus, ovary, brain, cartilage, and numerous other tissues, as well as chondroblastoma cell lines.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: a disintegrin and metalloproteinase; A disintegrin and metalloproteinase with thrombospondin motifs 11; A disintegrin and metalloproteinase with thrombospondin motifs 5; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5; a disintegrin-like and metalloprotease (reprolysin type) with thrombospondin type 1 motif, 5 (aggrecanase-2); ADAM; ADAM-TS 11; ADAM-TS 5; ADAM-TS5; ADAMs; ADAMTS family member; ADAMTS-5; ADAMTS11; ADAMTS family aggrecanase; ADMP-2; Aggrecanase 2; Aggrecanase-2; metalloendopeptidases

          View more View less

          Gene Aliases: ADAM-TS 11; ADAM-TS 5; ADAM-TS5; ADAMTS-11; ADAMTS-5; ADAMTS11; ADAMTS5; ADMP-2; ADMP2

          View more View less

          UniProt ID: (Human) Q9UNA0

          View more View less

          Entrez Gene ID: (Human) 11096, (Pig) 100522952, (Bovine) 286805

          View more View less

          Function(s)
          endopeptidase activity metalloendopeptidase activity integrin binding protein binding heparin binding peptidase activity metallopeptidase activity zinc ion binding hydrolase activity identical protein binding metal ion binding extracellular matrix binding metalloprotease
          Process(es)
          aortic valve morphogenesis pulmonary valve morphogenesis endocardial cushion morphogenesis proteolysis myoblast fusion extracellular matrix disassembly extracellular matrix organization defense response to bacterium negative regulation of cold-induced thermogenesis
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-d4j86:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline