Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • CENTG1 Antibodies

          Proteintech

          AGAP2 Polyclonal Antibody

          1 Reference
          View all (10) CENTG1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite AGAP2 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (7)
          AGAP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          AGAP2 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 7

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AGAP2 Antibody (14690-1-AP) in ICC/IF

          Immunofluorescent analysis of Hela cells, using AGAP2 antibody 14690-1-AP at 1:50 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). Blue pseudocolor = DAPI (fluorescent DNA dye). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AGAP2 Antibody in Immunocytochemistry (ICC/IF)
          AGAP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AGAP2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AGAP2 Antibody in Western Blot (WB)
          AGAP2 Antibody in Western Blot (WB)
          AGAP2 Antibody in Western Blot (WB)
          AGAP2 Antibody in Immunoprecipitation (IP)
          AGAP2 Polyclonal Antibody

          Product Details

          14690-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          AGAP2 Fusion Protein Ag6142 (848-1192 aa encoded by BC028020)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.17 mg/mL

          Amount

          25.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MVKKQRRKK LTTPSKTEGS AGQAEEENFE FLIVSSTGQT WHFEAASFEE RDAWVQAIES QILASLQCCE SSKVKLRTDS QSEAVAIQAI RNAKGNSICV DCGAPNPTWA SLNLGALICI ECSGIHRNLG THLSRVRSLD LDDWPRELTL VLTAIGNDTA NRVWESDTRG RAKPSRDSSR EERESWIRAK YEQLLFLAPL STSEEPLGRQ LWAAVQAQDV ATVLLLLAHA RHGPLDTSVE DPQLRSPLHL AAELAHVVIT QLLLWYGADV AARDAQGRTA LFYARQAGSQ LCADILLQHG CPGEGGSAAT TPSAATTPSI TATPSPRRRS SAASVGRADA PVALV (848-1192 aa encoded by BC028020)

          Target Information

          The protein encoded by this gene belongs to the centaurin gamma-like family. It mediates anti-apoptotic effects of nerve growth factor by activating nuclear phosphoinositide 3-kinase. It is overexpressed in cancer cells, and promotes cancer cell invasion. Alternatively spliced transcript variants encoding different isoforms have been described for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: AGAP-2; Arf GAP with GTP-binding protein-like, ANK repeat and PH domains 2; Arf-GAP with GTPase, ANK repeat and PH domain-containing protein 2; centaurin gamma1; centaurin, gamma 1; Centaurin-gamma-1; Cnt-g1; FLJ16430; GGAP2; GTP-binding and GTPase activating protein 2; GTP-binding and GTPase-activating protein 2; nuclear GTPase PIKE; Phosphatidylinositol 3-kinase enhancer; phosphatidylinositol-3-kinase enhancer; phosphoinositide 3-kinase enhancer; PIKE; PIKE-L; unnamed protein product

          View more View less

          Gene Aliases: AGAP-2; AGAP2; CENTG1; cnt-g1; GGAP2; KIAA0167; mKIAA0167; PIKE

          View more View less

          UniProt ID: (Human) Q99490, (Mouse) Q3UHD9, (Rat) Q8CGU4

          View more View less

          Entrez Gene ID: (Human) 116986, (Mouse) 216439, (Rat) 65218

          View more View less

          Function(s)
          nucleotide binding GTPase activity GTPase activator activity protein binding ATP binding GTP binding zinc ion binding protein kinase binding protein kinase activator activity phosphatidylinositol 3-kinase regulator activity metal ion binding phosphatidylinositol 3-kinase activator activity
          Process(es)
          small GTPase mediated signal transduction positive regulation of phosphatidylinositol 3-kinase signaling endosomal transport actin cytoskeleton organization positive regulation of mammary gland epithelial cell proliferation negative regulation of protein catabolic process negative regulation of apoptotic process negative regulation of neuron apoptotic process positive regulation of GTPase activity regulation of phosphatidylinositol 3-kinase activity positive regulation of protein kinase activity positive regulation of transcription from RNA polymerase II promoter positive regulation of JAK-STAT cascade mammary gland alveolus development negative regulation of extrinsic apoptotic signaling pathway in absence of ligand protein transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline