FIGURE: 1 / 3
Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
As a component of the inner core of AMPAR complex, modifies AMPA receptor (AMPAR) gating.
Protein Aliases: Germ cell-specific gene 1-like protein; GSG1-like; GSG1-like protein; KTSR5831; unnamed protein product
Gene Aliases: AI852434; C230098I05Rik; g1l; G630023A01Rik; Gm637; GSG1L; PRO19651; RGD1562278; UNQ5831/PRO19651
UniProt ID: (Human) Q6UXU4, (Mouse) D3Z7H4, (Rat) D3ZK93
Entrez Gene ID: (Human) 146395, (Mouse) 269994, (Rat) 499263