    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • QRFPR Antibodies
          Alomone Labs, LTD
          QRFPR/GPR103 (extracellular) Polyclonal Antibody
          View all (9) QRFPR antibodies
          Tech Support
          Cite QRFPR/GPR103 (extracellular) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          QRFPR/GPR103 (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          QRFPR/GPR103 (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          QRFPR/GPR103 (extracellular) Antibody (AGR-033-50UL) in WB
          Western blot analysis of rat brain membranes (lanes 1 and 3), mouse brain membranes (lanes 2 and 4) and human LNCaP cell lysate (lanes 3 and 6): - 1-3. Anti-QRFPR/GPR103 (extracellular) Antibody (#AGR-033), (1:500).4-6. Anti-QRFPR/GPR103 (extracellular) Antibody , preincubated with QRFPR/GPR103 (extracellular) Blocking Peptide (#BLP-GR033). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          QRFPR/GPR103 (extracellular) Antibody in Western Blot (WB)
          QRFPR/GPR103 (extracellular) Polyclonal Antibody
          Product Details
          AGR-033-50UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)HNLTREQFIHRYGLR, corresponding to amino acid residues 18-32 of rat QRFP receptor, Extracellular, N-terminus
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          40 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          G protein-coupled receptors (GPRs) are a protein family of transmembrane receptors that transmit an extracellular signal (ligand binding) into an intracellular signal (G protein activation). GPR signaling is an evolutionarily ancient mechanism used by all eukaryotes to sense environmental stimuli and mediate cell-cell communication. All of the receptors have seven membrane-spanning domains and the extracellular parts of the receptor can be glycosylated. These extracellular loops also contain two highly conserved cysteine residues which create disulfide bonds to stabilize the receptor structure. GPR103 is a 455 amino acid protein with highest expression in the brain, retina, trigeminal ganglion, hypothalamus and vestibular nucleus. In peripheral tissues, GPR103 is expressed only in the heart, kidney and testis. GPR103 may regulate adrenal function. A hypothalamic neuropeptide of the RFamide family (26RFa) acts as an endogenous ligand for GPR103.
          Bioinformatics
          Protein Aliases: AQ27; G protein-coupled receptor 103; G-protein coupled receptor 103; Orexigenic neuropeptide QRFP receptor; peptide P518 receptor; Pyroglutamylated RF-amide peptide receptor; pyroglutamylated RFamide peptide receptor; QRFP receptor; SP9155
          View more View less
          Gene Aliases: AQ27; GPR103; QRFPR; SP9155
          View more View less
          UniProt ID: (Human) Q96P65, (Rat) P83858, (Mouse) P83861
          View more View less
          Entrez Gene ID: (Human) 84109, (Rat) 310327, (Mouse) 229214
          View more View less
          Function(s)
          G-protein coupled receptor activity neuropeptide Y receptor activity protein binding peptide binding signal transducer activity G-protein coupled receptor
          Process(es)
          signal transduction G-protein coupled receptor signaling pathway neuropeptide signaling pathway cellular response to hormone stimulus response to peptide
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline