    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
          • Primary Antibodies ›
          • GlyT1 Antibodies
          Alomone Labs, LTD
          GlyT1 (SLC6A9) (extracellular) Polyclonal Antibody
          View all (6) GlyT1 antibodies
          Tech Support
          Cite GlyT1 (SLC6A9) (extracellular) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          GlyT1 (SLC6A9) (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          GlyT1 (SLC6A9) (extracellular) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          GlyT1 (SLC6A9) (extracellular) Antibody (AGT-011-25UL) in WB
          Western blot analysis of rat brain (lanes 1 and 4), mouse brain (lanes 2 and 5) and human U87-MG glioma cell (lanes 3 and 6) lysates: - 1-3. Anti-GlyT1 (SLC6A9) (extracellular) Antibody (#AGT-011), (1:200).4-6. Anti-GlyT1 (SLC6A9) (extracellular) Antibody , preincubated with GlyT1/SLC6A9 (extracellular) Blocking Peptide (#BLP-GT011). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GlyT1 (SLC6A9) (extracellular) Antibody in Western Blot (WB)
          GlyT1 (SLC6A9) (extracellular) Polyclonal Antibody
          Product Details
          AGT-011-25UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)RLYVLKLSDDIGD, corresponding to amino acid residues 202-214 of rat GlyT1, 2nd extracellular loop
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          20 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          Slc6a9 is a glycine transporter (GlyTs) that belong to a family of high affinity Na+- and Cl-dependent neurotransmitter transporters proteins. There are two subtypes of glycine transporters; GlyT1 (Slc6a9) and GlyT2. Slc6a9 are predominantly located on glia within caudal brain areas. The amino acid glycine acts as an inhibitory neurotransmitter in the central nervous system. The protein encoded by the Slc6a9 gene is one of two transporters that stop glycine signaling by removing it from the synaptic cleft. Diseases associated with SLC6A9 include Glycine Encephalopathy With Normal Serum Glycine and Infantile Glycine Encephalopathy.
          Bioinformatics
          Protein Aliases: glycine transporter 1; glycine transporter variant 1a; GlyT-1; Sodium- and chloride-dependent glycine transporter 1; solute carrier family 6 (neurotransmitter transporter, glycine), member 9; Solute carrier family 6 member 9; unnamed protein product
          View more View less
          Gene Aliases: GCENSG; GLYT-1; GLYT-1b; GLYT1; SLC6A9
          View more View less
          UniProt ID: (Human) P48067, (Mouse) P28571, (Rat) P28572
          View more View less
          Entrez Gene ID: (Human) 6536, (Mouse) 14664, (Rat) 116509
          View more View less
          Function(s)
          sodium:amino acid symporter activity glycine transmembrane transporter activity symporter activity glycine:sodium symporter activity transmembrane transporter activity neurotransmitter:sodium symporter activity protein binding primary active transporter
          Process(es)
          neurotransmitter uptake neurotransmitter transport amino acid transport glycine transport sodium ion transmembrane transport positive regulation of hemoglobin biosynthetic process regulation of synaptic transmission, glycinergic glycine secretion, neurotransmission positive regulation of heme biosynthetic process transport across blood-brain barrier glycine import across plasma membrane transport cation transmembrane transport L-alpha-amino acid transmembrane transport amino acid transmembrane transport
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          India flag icon
          India
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline