Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • AIM2 Antibodies

          Invitrogen

          AIM2 Polyclonal Antibody, eBioscience™

          Advanced Verification
          12 Published Figures
          12 References
          View all (23) AIM2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite AIM2 Polyclonal Antibody, eBioscience™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          • Published Figures (12)
          • Advanced Verification (2)
          AIM2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          AIM2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 15

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AIM2 Antibody (14-6008-93) in WB

          Cell lysates prepared from Jurkat cells (lane 1) and mouse macrophages left untreated (lane 2) or treated for 12 hr with IFN gamma and LPS (lane 3) were immunoblotted with a 1:1000 dilution of the Anti-AIM2 polyclonal antibody. Bands were visualized using Anti-Rabbit IgG HRP. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AIM2 Antibody in Western Blot (WB)
          AIM2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          AIM2 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          AIM2 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          AIM2 Antibody in Western Blot (WB)
          AIM2 Antibody in Western Blot (WB)
          AIM2 Antibody in Western Blot (WB)
          AIM2 Antibody in Western Blot (WB)
          AIM2 Antibody in Western Blot (WB)
          AIM2 Antibody in Western Blot (WB)
          AIM2 Antibody in Western Blot (WB)
          AIM2 Antibody in Western Blot (WB)
          AIM2 Antibody in Western Blot (WB)
          AIM2 Antibody
          AIM2 Antibody

          Product Details

          14-6008-93

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-Dependent
          View 7 publications 7 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Non-human primate, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Purification

          Affinity chromatography

          Storage buffer

          PBS with 50% glycerol, 1mg/mL BSA

          Contains

          0.05% sodium azide

          Storage conditions

          4°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_10670279

          Product Specific Information

          Description: This polyclonal antibody reacts with human, mouse, and rat AIM2 (also known as Antigen isolated from Immunoselected Melanoma 2). A member of the HIN-2000 family, this 40-kDa cytoplasmic protein is expressed in the spleen, small intestine, peripheral blood lymphocytes, and testis. Moreover, AIM2 is expressed in tumors of the neuroectodermis, breast, ovary, and colon. As a component of the inflammosome, AIM2 detects double-stranded viral and bacterial DNA present in the cytosol and initiates Caspase1 activation. Expression of AIM2 can be induced by interferons.

          Applications Reported: This polyclonal antibody has been reported for use in immunoblotting (WB).

          Applications Tested: This polyclonal antibody has been tested by immunoblotting of Jurkat cells and mouse macrophages treated for 12 hrs with IFN gamma and LPS. This can be used at less than or equal to a 1:1000 dilution. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.

          Purity: Greater than 90%, as determined by SDS-PAGE.

          Aggregation: Less than 10%, as determined by HPLC.

          Filtration: 0.2 µm post-manufacturing filtered.

          Target Information

          AIM2 is a member of the IFI20X /IFI16 family. It plays a putative role in tumorigenic reversion and may control cell proliferation. Interferon-gamma induces expression of AIM2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Absent in melanoma 2; Ifi-210; Interferon-inducible protein 210; Interferon-inducible protein AIM2; Interferon-inducible protein p210; RP11-520H16.3; unnamed protein product

          View more View less

          Gene Aliases: AIM2; Gm1313; Ifi210; PYHIN4

          View more View less

          UniProt ID: (Human) O14862, (Mouse) Q91VJ1

          View more View less

          Entrez Gene ID: (Human) 9447, (Rat) 304987, (Mouse) 383619

          View more View less

          Function(s)
          DNA binding double-stranded DNA binding protein binding signaling adaptor activity pattern recognition receptor activity identical protein binding cysteine-type endopeptidase activator activity molecular_function DNA-binding transcription factor
          Process(es)
          activation of innate immune response pattern recognition receptor signaling pathway positive regulation of defense response to virus by host immune system process inflammatory response immune response cellular response to DNA damage stimulus brain development negative regulation of NF-kappaB transcription factor activity positive regulation of interleukin-1 beta production tumor necrosis factor-mediated signaling pathway cellular response to interferon-beta T cell homeostasis innate immune response positive regulation of inflammatory response regulation of behavior positive regulation of NF-kappaB transcription factor activity protein maturation defense response to virus negative regulation of protein kinase B signaling neuron cellular homeostasis pyroptosis cellular response to cytokine stimulus cellular response to xenobiotic stimulus AIM2 inflammasome complex assembly pyroptotic cell death pyroptosome complex assembly biological_process positive regulation of protein oligomerization cellular response to drug interleukin-1 beta secretion positive regulation of interleukin-1 beta secretion positive regulation of cysteine-type endopeptidase activity apoptotic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
          git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.49.0-Offline