Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • AKAP1 Antibodies

          Abnova

          AKAP1 Polyclonal Antibody, MaxPab™

          View all (10) AKAP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite AKAP1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          AKAP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          AKAP1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AKAP1 Antibody (H00008165-B01P) in WB

          Western Blot analysis of AKAP1 expression in transfected 293T cell line by AKAP1 MaxPab polyclonal antibody. Lane 1: AKAP1 transfected lysate(99.33 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AKAP1 Antibody in Western Blot (WB)

          Product Details

          H00008165-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          AKAP1 (AAH00729.1, 1 a.a. approximately 903 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAIQFRSLFP LALPGMLALL GWWWFFSRKK GHVSSHDEQQ VEAGAVQLRA DPAIKEPLPV EDVCPKVVST PPSVTEPPEK ELSTVSKLPA EPPALLQTHP PCRRSESSGI LPNTTDMRLR PGTRRDDSTK LELALTGGEA KSIPLECPLS SPKGVLFSSK SAEVCKQDSP FSRVPRKVQP GYPVVPAEKR SSGERARETG GAEGTGDAVL GEKVLEEALL SREHVLELEN SKGPSLASLE GEEDKGKSSS SQVVGPVQEE EYVAEKLPSR FIESAHTELA KDDAAPAPPV ADAKAQDRGV EGELGNEESL DRNEEGLDRN EEGLDRNEES LDRNEEGLDR NEEIKRAAFQ IISQVISEAT EQVLATTVGK VAGRVCQASQ LQGQKEESCV PVHQKTVLGP DTAEPATAEA AVAPPDAGLP LPGLPAEGSP PPKTYVSCLK SLLSSPTKDS KPNISAHHIS LASCLALTTP SEELPDRAGI LVEDATCVTC MSDSSQSVPL VASPGHCSDS FSTSGLEDSC TETSSSPRDK AITPPLPEST VPFSNGVLKG ELSDLGAEDG WTMDAEADHS GGSDRNSMDS VDSCCSLKKT ESFQNAQAGS NPKKVDLIIW EIEVPKHLVG RLIGKQGRYV SFLKQTSGAK IYISTLPYTQ SVQICHIEGS QHHVDKALNL IGKKFKELNL TNIYAPPLPS LALPSLPMTS WLMLPDGITV EVIVVNQVNA GHLFVQQHTH PTFHALRSLD QQMYLCYSQP GIPTLPTPVE ITVICAAPGA DGAWWRAQVV ASYEETNEVE IRYVHYGGYK RVKVDVLRQI RSDFVTLPFQ GAEVLLDSVM PLSDDDQFSP EADAAMSEMT GNTALLAQVT SYSPTGLPLI QLWSVVGDEV VLINRSLVER GLAQWVDSYY TSL

          Target Information

          This gene encodes an enzyme which reversibly catalyzes the phosphorolysis of purine nucleosides. The enzyme is trimeric, containing three identical subunits. Mutations which result in nucleoside phosphorylase deficiency result in defective T-cell (cell-mediated) immunity but can also affect B-cell immunity and antibody responses. Neurologic disorders may also be apparent in patients with immune defects. A known polymorphism at aa position 51 that does not affect enzyme activity has been described. A pseudogene has been identified on chromosome 2.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: A kinase (PRKA) anchor protein 1; A-kinase anchor protein 1, mitochondrial; A-kinase anchor protein 149 kDa; AKAP 149; D-AKAP-1; Dual specificity A-kinase-anchoring protein 1; dual-specificity A-kinase anchoring protein 1; PRKA1; protein kinase A anchoring protein 1; Protein kinase A-anchoring protein 1; protein kinase A1; protein phosphatase 1, regulatory subunit 43; S-AKAP84; spermatid A-kinase; Spermatid A-kinase anchor protein 84; testicular secretory protein Li 5; tudor domain containing 17; unnamed protein product

          View more View less

          Gene Aliases: AKAP; AKAP1; AKAP121; AKAP149; AKAP84; D-AKAP1; PPP1R43; PRKA1; SAKAP84; TDRD17

          View more View less

          UniProt ID: (Human) Q92667

          View more View less

          Entrez Gene ID: (Human) 8165

          View more View less

          Function(s)
          nucleic acid binding RNA binding protein binding protein kinase A regulatory subunit binding
          Process(es)
          apoptotic process antiviral innate immune response
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline