Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • ALAS1 Antibodies

          Invitrogen

          ALAS1 Polyclonal Antibody

          View all (16) ALAS1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ALAS1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          ALAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ALAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ALAS1 Antibody (PA1-41152) in IHC (P)

          Immunohistochemistry analysis of ALAS1 in human skin. Samples were incubated with ALAS1 polyclonal antibody (Product # PA1-41152) using a dilution of 5 µg/mL. Heat induced antigen retrieval in pH 6.0 citrate buffer was used for unmasking. A biotinylated secondary antibody followed by alkaline phosphatase-streptavidin was and chromogen was used for detection. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ALAS1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ALAS1 Antibody in Western Blot (WB)
          ALAS1 Antibody in Western Blot (WB)
          ALAS1 Polyclonal Antibody

          Product Details

          PA1-41152

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-3 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A portion of amino acids 480-530 of human ALAS-1.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein G

          Storage buffer

          PBS with 0.05% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_1070357

          Product Specific Information

          Suggested positive control: HepG2.

          Immunogen sequence homology: 100% homologous in human, chimp and beluga whale; 94% in dog and chicken; 88% in mouse, rat and lungfish; 83% in bovine, xenopus, pufferfish; 77% in coelacanth.

          Target Information

          ALAS-1 is a nuclear-encoded mitochondrial matrix enzyme which catalyzes the condensation of glycine with succinyl-CoA to form delta-amino levulinate, CO2 and CoA. It regulates the first and rate-limiting step of heme biosynthetic pathway. It is one of the two isoforms of ALAS and is a pyridoxal-5' phosphate dependant housekeeping enzyme. It is ubiquitously expressed and is responsible of providing heme for cytochromes and other hemoproteins. ALAS1 in liver undergoes negative feedback regulation by heme. Transcription of ALAS1 gene is stimulated by cAMP and respiratory uncoupling whereas Phorbol esters and insulin repress ALAS1 gene expression. ALAS1 is suggested to be involved in reciprocal regulation of Heme biosynthesis and circadian clock suggesting a potential target for treatment of circadian disorders. ALAS1 gene expression is down regulated in Acute Liver failure resulting in altered heme metabolism and liver function.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 5-aminolevulinate synthase 1; 5-aminolevulinate synthase, non-specific, mitochondrial; 5-aminolevulinate synthase, nonspecific, mitochondrial; 5-aminolevulinic acid synthase 1; ALA-S; ALAS-H; ALAS-N; aminolevulinate, delta-, synthase 1; aminolevulinic acid synthase 1; Delta-ALA synthase 1; delta-ALA synthetase; Delta-aminolevulinate synthase 1; MIG4; migration-inducing protein 4; unnamed protein product

          View more View less

          Gene Aliases: ALAS; ALAS-H; ALAS1; ALAS3; ALASH; MIG4; OK/SW-cl.121

          View more View less

          UniProt ID: (Human) P13196, (Rat) P13195

          View more View less

          Entrez Gene ID: (Human) 211, (Rat) 65155

          View more View less

          Function(s)
          5-aminolevulinate synthase activity protein binding transferase activity transferase activity, transferring acyl groups pyridoxal phosphate binding identical protein binding transaminase
          Process(es)
          response to hypoxia porphyrin-containing compound metabolic process protoporphyrinogen IX biosynthetic process heme biosynthetic process heme a biosynthetic process heme B biosynthetic process response to xenobiotic stimulus response to herbicide response to nickel cation response to nutrient levels response to cobalt ion cellular response to insulin stimulus tetrapyrrole biosynthetic process response to gonadotropin hemoglobin biosynthetic process response to ethanol heme O biosynthetic process erythrocyte development response to cAMP response to platinum ion response to bile acid response to organic substance response to organic cyclic compound response to drug cellular response to organic cyclic compound
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-vzwvz:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline