Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LGVVLVIGT WNYPFNLTIQ PMVGAIAAGN AVVLKPSELS ENMASLLATI IPQYLDKDLY PVINGGVPET TELLKERFDH ILYTGSTGVG KIIMTAAAKH LTPVTLELGG KSPCYVDKNC DLDVACRRIA WGKFMNSGQT CVAPDYILCD PSIQNQIVEK LKKSLKEFYG EDAKKSRDYG RIISARHFQR VMGLIEGQKV AYGGTGDAAT RYIAPTILTD VDPQSPVMQE EIFGPVLPIV CVRSLEEAIQ FINQREKPLA LYMFSSNDKV IKKMIAETSS GGVAANDVIV HITLHSLPFG GVGNSGMGSY HGKKSFETFS HRRSCLVRPL MNDEGLKVRY PPSPAKMTQH (105-453 aa encoded by BC004370)
Aldh3A1 is a member of the aldehyde dehydrogenase superfamily, a group of NAD(P)(+)-dependent enzymes that catalyze the oxidation of a wide spectrum of aliphatic and aromatic aldehydes. Aldh3A1 is highly expressed in stomach and even more strongly in cornea, representing between 5 to 50% of the water soluble protein fraction in mammalian corneas. It is thought that Aldh3A1 acts to protect the cornea from UV-induced oxidative stress by not only detoxification of reactive aldehydes by also through the direct absorption of UV energy. However, corneas from Aldh3A1-null mice are indistinguishable from those from wild-type mice; mice lacking both Aldh3A1 and Aldh1A1 showed increased cataract formation following UVB exposure, suggesting that Aldh1A1 may be able to compensate for the loss of Aldh3A1.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alcohol dehydrogenase family 3, subfamily A1; aldehyde dehydrogenase; Aldehyde dehydrogenase (Ahd-c); aldehyde dehydrogenase 3, stomach cytosolic (class 3); aldehyde dehydrogenase 3A1; aldehyde dehydrogenase 4; aldehyde dehydrogenase class 3; aldehyde dehydrogenase family 3 member A1; aldehyde dehydrogenase family 3 subfamily A1; aldehyde dehydrogenase isozyme 3; aldehyde dehydrogenase type III; dioxin-inducible aldehyde dehydrogenase 3; HTC-ALDH; MGC10406; stomach aldehyde dehydrogenase; tumor-associated aldehyde dehydrogenase tumor ALDH; unnamed protein product
Gene Aliases: Ahd-4; Ahd-c; Ahd4; AHDC; Aldh; ALDH3; ALDHIII
UniProt ID: (Human) P30838, (Mouse) P47739, (Rat) P11883
Entrez Gene ID: (Human) 218, (Mouse) 11670, (Rat) 25375
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support