Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MAAAAAETP EVLRECGCKG IRTCLICERQ RGSDPPWELP PAKTYRFIYC SDTGWAVGTE ESDFEGWAFP FPGVMLIEDF VTREEEAELV RLMDRDPWKL SQSGRRKQDY GPKVNFRKQK LKTEGFCGLP SFSREVVRRM GLYPGLEGFR PVEQCNLDYC PERGSAIDPH LDDAWLWGER LVSLNLLSPT VLSMCREAPG SLLLCSAPSA APEALVDSVI APSRSVLCQE VEVAIPLPAR SLLVLTGAAR HQWKHAIHRR HIEARRVCVT FRELSAEFGP GGRQQELGQE LLRIALSFQG RPV (1-302 aa encoded by BC002820)
Dioxygenase that mediates demethylation of actin monomethylated at 'Lys-84' (K84me1), thereby acting as a regulator of actomyosin-processes (PubMed:23673617). Demethylation of actin K84me1 is required for maintaining actomyosin dynamics supporting normal cleavage furrow ingression during cytokinesis and cell migration (PubMed:23673617). In addition to proteins, also demethylates DNA: specifically demethylates DNA methylated on the 6th position of adenine (N(6)-methyladenosine) DNA, thereby regulating Polycomb silencing. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alkB homolog 4, lysine demthylase; alkB, alkylation repair homolog 4; alkylated DNA repair protein alkB homolog 4; DNA N6-methyl adenine demethylase ALKBH4; lysine-specific demethylase ALKBH4; probable alpha-ketoglutarate-dependent dioxygenase ABH4; unnamed protein product
Gene Aliases: 2010004B12Rik; ABH4; Abj4; RGD1308608
UniProt ID: (Human) Q9NXW9, (Mouse) Q9D8F1
Entrez Gene ID: (Human) 54784, (Mouse) 72041, (Rat) 288587
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support