Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • ALOXE3 Antibodies

          Abnova

          ALOXE3 Polyclonal Antibody, MaxPab™

          View all (6) ALOXE3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ALOXE3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ALOXE3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ALOXE3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ALOXE3 Antibody (H00059344-B01P) in WB

          Western Blot analysis of ALOXE3 expression in transfected 293T cell line by ALOXE3 MaxPab polyclonal antibody. Lane 1: ALOXE3 transfected lysate(78.21 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ALOXE3 Antibody in Western Blot (WB)
          ALOXE3 Polyclonal Antibody, MaxPab™

          Product Details

          H00059344-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ALOXE3 (AAI01939.1, 1 a.a. approximately 711 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAVYRLCVTT GPYLRAGTLD NISVTLVGTC GESPKQRLDR MGRDFAPGSV QKYKVRCTAE LGELLLLRVH KERYAFFRKD SWYCSRICVT EPDGSVSHFP CYQWIEGYCT VELRPGTART ICQDSLPLLL DHRTRELRAR QECYRWKIYA PGFPCMVDVN SFQEMESDKK FALTKTTTCV DQGDSSGNRY LPGFPMKIDI PSLMYMEPNV RYSATKTISL LFNAIPASLG MKLRGLLDRK GSWKKLDDMQ NIFWCHKTFT TKYVTEHWCE DHFFGYQYLN GVNPVMLHCI SSLPSKLPVT NDMVAPLLGQ DTCLQTELER GNIFLADYWI LAEAPTHCLN GRQQYVAAPL CLLWLSPQGA LVPLAIQLSQ TPGPDSPIFL PTDSEWDWLL AKTWVRNSEF LVHENNTHFL CTHLLCEAFA MATLRQLPLC HPIYKLLLPH TRYTLQVNTI ARATLLNPEG LVDQVTSIGR QGLIYLMSTG LAHFTYTNFC LPDSLRARGV LAIPNYHYRD DGLKIWAAIE SFVSEIVGYY YPSDASVQQD SELQAWTGEI FAQAFLGRES SGFPSRLCTP GEMVKFLTAI IFNCSAQHAA VNSGQHDFGA WMPNAPSSMR QPPPQTKGTT TLKTYLDTLP EVNISCNNLL LFWLVSQEPK DQRPLGTYPD EHFTEEAPRR SIAAFQSRLA QISRDIQERN QGLALPYTYL DPPLIENSVS I

          Target Information

          This gene is a member of the lipoxygenase family, which are catabolized by arachidonic acid-derived compounds. The encoded enzyme is a hydroperoxide isomerase that synthesizes a unique type of epoxy alcohol (8R-hydroxy-11R,12R-epoxyeicosa-5Z,9E,14Z-trienoic acid) from 12R-hydroperoxyeicosatetraenoic acid (12R-HPETE). This epoxy alcohol can activate the the nuclear receptor peroxisome proliferator-activated receptor alpha (PPARalpha), which is implicated in epidermal differentiation. Loss of function of the enzyme encoded by this gene results in ichthyosis, implicating the function of this gene in the differentiation of human skin. This gene is part of a cluster of lipoxygenase genes on 17p13.1. Mutations in this gene result in nonbullous congenital ichthyosiform erythroderma (NCIE). Multiple transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Sep 2009].

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: arachidonate lipoxygenase 3; e-LOX-3; epidermal lipoxygenase; Epidermal lipoxygenase-3; Epidermal LOX-3; Epidermis-type lipoxygenase 3; Hydroperoxide isomerase ALOXE3; Hydroperoxy dehydratase ALOXE3; Hydroperoxy icosatetraenoate dehydratase; Hydroperoxy icosatetraenoate isomerase; Lamellar ichthyosis 5; unnamed protein product

          View more View less

          Gene Aliases: ALOXE3; ARCI3; E-LOX; eLOX-3; eLOX3; LI5

          View more View less

          UniProt ID: (Human) Q9BYJ1

          View more View less

          Entrez Gene ID: (Human) 59344

          View more View less

          Function(s)
          iron ion binding protein binding oxidoreductase activity oxidoreductase activity, acting on single donors with incorporation of molecular oxygen, incorporation of two atoms of oxygen lyase activity isomerase activity metal ion binding intramolecular transferase activity, transferring hydroxy groups dioxygenase activity hydroperoxy icosatetraenoate isomerase activity hydroperoxy icosatetraenoate dehydratase activity
          Process(es)
          lipid metabolic process fatty acid metabolic process sphingolipid metabolic process sensory perception of pain arachidonic acid metabolic process lipoxygenase pathway lipid oxidation peroxisome proliferator activated receptor signaling pathway linoleic acid metabolic process fat cell differentiation ceramide biosynthetic process hepoxilin biosynthetic process establishment of skin barrier
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline