Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • Alsin Antibodies

          Abnova

          ALS2 Polyclonal Antibody, MaxPab™

          View all (12) Alsin antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ALS2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ALS2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ALS2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ALS2 Antibody (H00057679-D01P) in WB

          Western Blot analysis of ALS2 expression in transfected 293T cell line (H00057679-T02) by ALS2 MaxPab polyclonal antibody.

          Lane 1: ALS2 transfected lysate(135.70 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ALS2 Antibody in Western Blot (WB)
          ALS2 Polyclonal Antibody, MaxPab™

          Product Details

          H00057679-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ALS2 (AAH71576.1, 1 a.a. approximately 1237 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MDSKKRSSTE AEGSKERGLV HIWQAGSFPI TPERLPGWGG KTVLQAALGV KHGVLLTEDG EVYSFGTLPW RSGPVEICPS SPILENALVG QYVITVATGS FHSGAVTDNG VAYMWGENSA GQCAVANQQY VPEPNPVSIA DSEASPLLAV RILQLACGEE HTLALSISRE IWAWGTGCQL GLITTAFPVT KPQKVEHLAG RVVLQVACGA FHSLALVQCL PSQDLKPVPE RCNQCSQLLI TMTDKEDHVI ISDSHCCPLG VTLTESQAEN HASTALSPST ETLDRQEEVF ENTLVANDQS VATELNAVSA QITSSDAMSS QQNVMGTTEI SSARNIPSYP DTQAVNEYLR KLSDHSVRED SEHGEKPVPS QPLLEEAIPN LHSPPTTSTS ALNSLVVSCA SAVGVRVAAT YEAGALSLKK VMNFYSTTPC ETGAQAGSSA IGPEGLKDSR EEQVKQESMQ GKKSSSLVDI REEETEGGSR RLSLPGLLSQ VSPRLLRKAA RVKTRTVVLT PTYSGEADAL LPSLRIEVWT WGKGKEGQLG HGDVLPRLQP LCVKCLDGKE VIHLEAGGYH SLALTAKSQV YSWGSNTFGQ LGHSDFPTTV PRLAKISSEN GVWSIAAGRD YSLFLVDTED FQPGLYYSGR QDPTEGDNLP ENHSGSKTPV LLSCSKLGYI SRVTAGKDSY LALVDKNIMG YIASLHELAT TERRFYSKLS DIKSQILRPL LSLENLGTTT TVQLLQEVAS RFSKLCYLIG QHGASLSSFL HGVKEARSLV ILKHSSLFLD SYTEYCTSIT NFLVMGGFQL LAKPAIDFLN KNQELLQDLS EVNDENTQLM EILNTLFFLP IRRLHNYAKV LLKLATCFEV ASPEYQKLQD SSSCYECLAL HLGRKRKEAE YTLGFWKTFP GKMTDSLRKP ERRLLCESSN RALSLQHAGR FSVNWFILFN DALVHAQFST HHVFPLATLW AEPLSEEAGG VNGLKITTPE EQFTLISSTP QEKTKWLRAI SQAVDQALRG MSDLPPYGSG SSVQRQEPPI SRSAKYTFYK DPRLKDATYD GRWLSGKPHG RGVLKWPDGK MYSGMFRNGL EDGYGEYRIP NKAMNKEDHY VGHWKEGKMC GQGVYSYASG EVFEGCFQDN MRHGHGLLRS GKLTSSSPSM FIGQWVMDKK AGYGVFDDIT RGEKYMGMWQ DDVCQGNGVV VTQFGLYYEG NFHLNKMMGN GVLLSEDDTI YEGEFSDDWT LSGKVGN

          Target Information

          The function of this protein remains unknown.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ALS2; ALS2, alsin Rho guanine nucleotide exchange factor; Alsin; amyotrophic lateral sclerosis 2 (juvenile); amyotrophic lateral sclerosis 2 (juvenile) isoform ALS2_1; amyotrophic lateral sclerosis 2 (juvenile) isoform ALS2_2; Amyotrophic lateral sclerosis 2 chromosomal region candidate gene 6 protein; Amyotrophic lateral sclerosis 2 protein; FLJ31851; MGC87187; unnamed protein product

          View more View less

          Gene Aliases: ALS2; ALS2CR6; ALSJ; IAHSP; KIAA1563; PLSJ

          View more View less

          UniProt ID: (Human) Q96Q42

          View more View less

          Entrez Gene ID: (Human) 57679

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity GTPase activator activity protein binding small GTPase binding identical protein binding protein homodimerization activity protein serine/threonine kinase activator activity
          Process(es)
          behavioral fear response receptor recycling response to oxidative stress endosome organization lysosomal transport neuromuscular junction development locomotory behavior protein localization vesicle organization endosomal transport positive regulation of Rac protein signal transduction synaptic transmission, glutamatergic positive regulation of GTPase activity positive regulation of protein kinase activity neuron projection morphogenesis regulation of endosome size protein homooligomerization regulation of biological quality regulation of postsynaptic membrane neurotransmitter receptor levels
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline