Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          • Primary Antibodies ›
          • ALS2CL Antibodies

          Abnova

          ALS2CL Polyclonal Antibody, MaxPab™

          View all (3) ALS2CL antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ALS2CL Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ALS2CL Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ALS2CL Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ALS2CL Antibody (H00259173-B01P) in WB

          Western Blot analysis of ALS2CL expression in transfected 293T cell line by ALS2CL MaxPab polyclonal antibody. Lane 1: ALS2CL transfected lysate(104.83 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ALS2CL Antibody in Western Blot (WB)
          ALS2CL Polyclonal Antibody, MaxPab™

          Product Details

          H00259173-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ALS2CL (NP_667340.2, 1 a.a. approximately 953 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MCNPEEAALL RLEEVFSATL AHVNSLVLQP LLPAAPDPSD PWGRECLRLL QQLHKSSQQL WEVTEESLHS LQERLRYPDS TGLESLLLLR GADRVLQAHI EYIESYTSCM VVQAFQKAAK RRSEYWRGQR KALRQLLSGV SSEGSVGASL GQALHQPLAH HVQQYVLLLL SLGDTIGEHH PTRELVVNAV TLFGNLQSFM KQELDQAVAT QALWHTLRGR LRDVLCTPAH RLLQDSQDVP VTVAPLRAER VLLFDDALVL LQGHNVHTFD LKLVWVDPGQ DGCTFHLLTP EEEFSFCAKD SQGQAVWQWK VTWAVHQALH GKKDFPVLGA GLEPSQPPDC RCAEYTFQAE GRLCQATYEG EWCRGRPHGK GTLKWPDGRN HVGNFCQGLE HGFGIRLLPQ ASEDKFDCYK CHWREGSMCG YGICEYSTDE VYKGYFQEGL RHGFGVLESG PQAPQPFRYT GHWERGQRSG YGIEEDGDRG ERYIGMWQAG QRHGPGVMVT QAGVCYQGTF QADKTVGPGI LLSEDDSLYE GTFTRDLTLM GKGKVTFPNG FTLEGSFGSG AGRGLHTQGV LDTAALPPDP SSTCKRQLGV GAFPVESRWQ GVYSPFRDFV CAGCPRDLQE ALLGFDVQSS RELRRSQDYL SCERTHPEDS VGSMEDILEE LLQHREPKAL QLYLRKALSN SLHPLGKLLR TLMLTFQATY AGVGANKHLQ ELAQEEVKQH AQELWAAYRG LLRVALERKG QALEEDEDTE TRDLQVHGLV LPLMLPSFYS ELFTLYLLLH EREDSFYSQG IANLSLFPDT QLLEFLDVQK HLWPLKDLTL TSNQRYSLVR DKCFLSATEC LQKIMTTVDP REKLEVLERT YGEIEGTVSR VLGREYKLPM DDLLPLLIYV VSRARIQHLG AEIHLIRDMM DPNHTGGLYD FLLTALESCY EHIQKEDMRL HRLPGHWHSR ELW

          Target Information

          Acts as a guanine nucleotide exchange factor (GEF) for Rab5 GTPase. Regulates the ALS2-mediated endosome dynamics. [UniProt]

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: ALS2 C-terminal-like protein; unnamed protein product

          View more View less

          Gene Aliases: ALS2CL; RN49018

          View more View less

          UniProt ID: (Human) Q60I27

          View more View less

          Entrez Gene ID: (Human) 259173

          View more View less

          Function(s)
          guanyl-nucleotide exchange factor activity GTPase activator activity protein binding small GTPase binding
          Process(es)
          endosomal transport
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline