FIGURE: 1 / 2
PA1-31835 detects ALS2CR1 from human samples.
PA1-31835 has been successfully used in Western blot applications.
The PA1-31835 immunogen is: Peptide with sequence ILSETDRDPLQVV, from C Terminus of the protein sequence according to NP_068596
ALS2CR1 may function as a transcriptional corepressor through its interaction with COPS2, negatively regulating the expression of genes involved in neuronal differentiation.