Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • AMOTL1 Antibodies

          Abnova

          AMOTL1 Polyclonal Antibody, MaxPab™

          View all (6) AMOTL1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite AMOTL1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          AMOTL1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          AMOTL1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          AMOTL1 Antibody (H00154810-D01P) in WB

          Western Blot analysis of AMOTL1 expression in transfected 293T cell line by AMOTL1 MaxPab polyclonal antibody. Lane 1: AMOTL1 transfected lysate(106.60 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          AMOTL1 Antibody in Western Blot (WB)
          AMOTL1 Polyclonal Antibody, MaxPab™

          Product Details

          H00154810-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          AMOTL1 (NP_570899.1, 1 a.a. approximately 956 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MWRAKLRRGT CEPAVKGSPS ACYSPSSPVQ VLEDSTYFSP DFQLYSGRHE TSALTVEATS SIREKVVEDP LCNFHSPNFL RISEVEMRGS EDAAAGTVLQ RLIQEQLRYG TPTENMNLLA IQHQATGSAG PAHPTNNFSS TENLTQEDPQ MVYQSARQEP QGQEHQVDNT VMEKQVRSTQ PQQNNEELPT YEEAKAQSQF FRGQQQQQQQ QGAVGHGYYM AGGTSQKSRT EGRPTVNRAN SGQAHKDEAL KELKQGHVRS LSERIMQLSL ERNGAKQHLP GSGNGKGFKV GGGPSPAQPA GKVLDPRGPP PEYPFKTKQM MSPVSKTQEH GLFYGDQHPG MLHEMVKPYP APQPVRTDVA VLRYQPPPEY GVTSRPCQLP FPSTMQQHSP MSSQTSSASG PLHSVSLPLP LPMALGAPQP PPAASPSQQL GPDAFAIVER AQQMVEILTE ENRVLHQELQ GYYDNADKLH KFEKELQRIS EAYESLVKST TKRESLDKAM RNKLEGEIRR LHDFNRDLRD RLETANRQLS SREYEGHEDK AAEGHYASQN KEFLKEKEKL EMELAAVRTA SEDHRRHIEI LDQALSNAQA RVIKLEEELR EKQAYVEKVE KLQQALTQLQ SACEKREQME RRLRTWLERE LDALRTQQKH GNGQPANMPE YNAPALLELV REKEERILAL EADMTKWEQK YLEESTIRHF AMNAAATAAA ERDTTIINHS RNGSYGESSL EAHIWQEEEE VVQANRRCQD MEYTIKNLHA KIIEKDAMIK VLQQRSRKDA GKTDSSSLRP ARSVPSIAAA TGTHSRQTSL TSSQLAEEKK EEKTWKGSIG LLLGKEHHEH ASAPLLPPPP TSALSSIAST TAASSAHAKT GSKDSSTQTD KSAELFWPSM ASLPSRGRLS TTPAHSPVLK HPAAKGTAEK LENSPGHGKS PDHRGRVSSL LHKPEFPDGE MMEVLI

          Target Information

          The protein encoded by this gene is a peripheral membrane protein that is a component of tight junctions or TJs. TJs form an apical junctional structure and act to control paracellular permeability and maintain cell polarity. This protein is related to angiomotin, an angiostatin binding protein that regulates endothelial cell migration and capillary formation. Two transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Angiomotin-like protein 1; B-motin; JEAP; junction-enriched and -associated protein; junction-enriched and associated protein; unnamed protein product

          View more View less

          Gene Aliases: 2310010G08Rik; 2310067L22Rik; 4932416D09Rik; AMOTL1; CFCHS; JEAP; mFLJ00155

          View more View less

          UniProt ID: (Human) Q8IY63, (Mouse) Q9D4H4

          View more View less

          Entrez Gene ID: (Human) 154810, (Mouse) 75723

          View more View less

          Function(s)
          protein binding identical protein binding
          Process(es)
          establishment of cell polarity involved in ameboidal cell migration Wnt signaling pathway hippo signaling positive regulation of blood vessel endothelial cell migration angiogenesis actin cytoskeleton organization regulation of cell migration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline