    • See all product categories
  • Applications
    • Cell Analysis
    • Protein Biology
    • Lab Informatics
    • 360° CDMO and CRO Solutions
  • Help and Support
    • Customer Center
    • Contact Us
    • Certificates of Analysis and Conformance
    • Safety Data Sheets (SDS)
    • Manuals
    • How to Cite Our Products in a Paper
    • Instrument Support
    • Knowledge Base and Product FAQs
          • Primary Antibodies ›
          • Annexin A10 Antibodies
          Fabgennix
          Annexin A10 Polyclonal Antibody, Biotin
          View all (26) Annexin A10 antibodies
          Tech Support
          Cite Annexin A10 Polyclonal Antibody, Biotin
          Additional Information:
          {{banner.description}}
          Annexin A10 Polyclonal Antibody, Biotin
          Product Details
          ANXA10-BIOTIN
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:500-1:1,000
          -
          ELISA (ELISA)
          1:10,000
          -
          Immunoprecipitation (IP)
          1:50-1:250
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          Synthetic peptide taken within amino acid region 60-100 on human Annexin A10 protein.
          3D Epitope / Immunogen
          Conjugate
          Biotin Biotin Biotin
          Form
          Liquid
          Concentration
          0.5-1.5 mg/mL
          Purification
          Affinity chromatography
          Storage buffer
          proprietary buffer, pH 7.4-7.8, with 30% glycerol, 0.5% BSA
          Contains
          0.02% sodium azide
          Storage conditions
          -20°C
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The function of this gene has not yet been determined.
          Bioinformatics
          Protein Aliases: annexin 14; Annexin A10; Annexin-10; Annexin-14
          View more View less
          Gene Aliases: ANX14; ANXA10; RGD1560956
          View more View less
          UniProt ID: (Human) Q9UJ72
          View more View less
          Entrez Gene ID: (Human) 11199, (Rat) 498622
          View more View less
          Function(s)
          phosphatidylserine binding calcium ion binding protein binding calcium-dependent phospholipid binding calcium-binding protein
          • Trademarks
          • Fair Trade Fair Trade
          Korea flag icon
          Korea
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Customer Help Desk | working day 09:00~18:00
          1661-9555   |   Live Chat   |   카카오톡 상담
          Service Call Center | working day 09:00~18:00
          1661-5055   |   Live Chat
          Thermo Fisher Scientific Korea Ltd.
          Representative : Soojin Seok
          Company Registration No. : 117-81-46910
          Thermo Fisher Scientific Solutions LLC
          Representative : Soojin Seok
          Company Registration No. : 114-86-04783
          Location: 12F Suseo Office Building, 281 Gwangpyeong-ro, Gangnam-gu, Seoul, Korea(06349) | Mail-Order Business Registration : 2015-Seoul Gangnam-00898 | Payment : ShinHan Bank 140-004-396660 (Thermo Fisher Scientific Solutions LLC)
          ISMS Logo
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline