Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • APAF1 Antibodies

          Abnova

          APAF1 Polyclonal Antibody, MaxPab™

          View all (23) APAF1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite APAF1 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          APAF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          APAF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          APAF1 Antibody (H00000317-B01P) in WB

          Western Blot analysis of APAF1 expression in transfected 293T cell line by APAF1 MaxPab polyclonal antibody. Lane 1: APAF1 transfected lysate(132.55 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          APAF1 Antibody in Western Blot (WB)
          APAF1 Polyclonal Antibody, MaxPab™

          Product Details

          H00000317-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          APAF1 (NP_863658.1, 1 a.a. approximately 1205 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MDAKARNCLL QHREALEKDI KTSYIMDHMI SDGFLTISEE EKVRNEPTQQ QRAAMLIKMI LKKDNDSYVS FYNALLHEGY KDLAALLHDG IPVVSSSSGK DSVSGITSYV RTVLCEGGVP QRPVVFVTRK KLVNAIQQKL SKLKGEPGWV TIHGMAGCGK SVLAAEAVRD HSLLEGCFPG GVHWVSVGKQ DKSGLLMKLQ NLCTRLDQDE SFSQRLPLNI EEAKDRLRIL MLRKHPRSLL ILDDVWDSWV LKAFDSQCQI LLTTRDKSVT DSVMGPKYVV PVESSLGKEK GLEILSLFVN MKKADLPEQA HSIIKECKGS PLVVSLIGAL LRDFPNRWEY YLKQLQNKQF KRIRKSSSYD YEALDEAMSI SVEMLREDIK DYYTDLSILQ KDVKVPTKVL CILWDMETEE VEDILQEFVN KSLLFCDRNG KSFRYYLHDL QVDFLTEKNC SQLQDLHKKI ITQFQRYHQP HTLSPDQEDC MYWYNFLAYH MASAKMHKEL CALMFSLDWI KAKTELVGPA HLIHEFVEYR HILDEKDCAV SENFQEFLSL NGHLLGRQPF PNIVQLGLCE PETSEVYQQA KLQAKQEVDN GMLYLEWINK KNITNLSRLV VRPHTDAVYH ACFSEDGQRI ASCGADKTLQ VFKAETGEKL LEIKAHEDEV LCCAFSTDDR FIATCSVDKK VKIWNSMTGE LVHTYDEHSE QVNCCHFTNS SHHLLLATGS SDCFLKLWDL NQKECRNTMF GHTNSVNHCR FSPDDKLLAS CSADGTLKLW DATSANERKS INVKQFFLNL EDPQEDMEVI VKCCSWSADG ARIMVAAKNK IFLWNTDSRS KVADCRGHLS WVHGVMFSPD GSSFLTSSDD QTIRLWETKK VCKNSAVMLK QEVDVVFQEN EVMVLAVDHI RRLQLINGRT GQIDYLTEAQ VSCCCLSPHL QYIAFGDENG AIEILELVNN RIFQSRFQHK KTVWHIQFTA DEKTLISSSD DAEIQVWNWQ LDKCIFLRGH QETVKDFRLL KNSRLLSWSF DGTVKVWNII TGNKEKDFVC HQGTVLSCDI SHDATKFSST SADKTAKIWS FDLLLPLHEL RGHNGCVRCS AFSVDSTLLA TGDDNGEIRI WNVSNGELLH LCAPLSEEGA ATHGGWVTDL CFSPDGKMLI SAGGYIKWWN VVTGESSQTF YTNGTNLKKI HVSPDFKTYV TVDNLGILYI LQTLE

          Target Information

          APAF1 gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: APAF-1; Apaf-1XL; Apaf1 variant; Apoptotic protease-activating factor 1; cytochrome c and dATP-dependent procaspase-9 activation factor; DKFZp781B1145; KIAA0413; similar to C. elegans cell death gene ced-4

          View more View less

          Gene Aliases: APAF-1; APAF1; CED4; KIAA0413

          View more View less

          UniProt ID: (Human) O14727

          View more View less

          Entrez Gene ID: (Human) 317

          View more View less

          Function(s)
          nucleotide binding protein binding ATP binding cysteine-type endopeptidase activator activity involved in apoptotic process heat shock protein binding identical protein binding ADP binding cysteine-type endopeptidase activator activity
          Process(es)
          response to hypoxia kidney development neural tube closure apoptotic process nervous system development brain development response to nutrient cardiac muscle cell apoptotic process cell differentiation forebrain development regulation of apoptotic process positive regulation of apoptotic process system development neuron apoptotic process intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress cellular response to transforming growth factor beta stimulus response to G1 DNA damage checkpoint signaling apoptotic signaling pathway intrinsic apoptotic signaling pathway regulation of apoptotic DNA fragmentation positive regulation of apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-2r4kz:80/100.66.79.163:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline