    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • KCNMB2 Antibodies
          Alomone Labs, LTD
          slo beta 2 (KCNMB2) Polyclonal Antibody
          View all (10) KCNMB2 antibodies
          Tech Support
          Cite slo beta 2 (KCNMB2) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          slo beta 2 (KCNMB2) Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          slo beta 2 (KCNMB2) Antibody in Immunohistochemistry (IHC)
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          slo beta 2 (KCNMB2) Antibody (APC-034-25UL) in IHC
          Expression of KCNMB2in rat hippocampus - Immunohistochemical staining of rat hippocampus using Anti-slo beta 2 (KCNMB2) Antibody (#APC-034). A. KCNMB2appears in the pyramidal layer (green). B. Staining of interneurons with mouse Anti-parvalbumin (PV, red). C. Confocal merge of KCNMB2and PV demonstrates presence of KCNMB2also in PV (GABA containing) cells (orange). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          slo beta 2 (KCNMB2) Antibody in Immunohistochemistry (IHC)
          slo beta 2 (KCNMB2) Antibody in Western Blot (WB)
          slo beta 2 (KCNMB2) Polyclonal Antibody
          Product Details
          APC-034-25UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Immunohistochemistry (IHC)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)RHDEKRNIYQKIRDHDLLD, corresponding to amino acid residues 14-32 of humanKCNMB2, Intracellular, N-terminal domain
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          20 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which decreases the activation time of MaxiK alpha subunit currents. Alternative splicing results in multiple transcript variants of this gene. Additional variants are discussed in the literature, but their full length nature has not been described.
          Bioinformatics
          Protein Aliases: big potassium channel beta subunit 2; BK channel beta subunit 2; BK channel subunit beta-2; BKbeta2; calcium-activated potassium channel beta 2; Calcium-activated potassium channel subunit beta-2; Calcium-activated potassium channel, subfamily M subunit beta-2; Charybdotoxin receptor subunit beta-2; hbeta2; Hbeta3; hCG1646471; hKCNMB2; large conductance (BK) potassium channel beta2 subunit; K(VCA)beta-2; large conductance calcium-activated potassium channel beta 2 subunit; large-conductance Ca2+-activated K+ channel beta2 subunit; Maxi K channel subunit beta-2; MaxiK channel beta 2 subunit; MaxiK channel beta-subunit 2; modulatory subunit of the voltage and Ca2+ activated K+ (MaxiK) channel; potassium channel subfamily M regulatory beta subunit 2; potassium large conductance calcium-activated channel, subfamily M, beta member 2; Rbeta3; Slo-beta-2; Slo-beta2; unnamed protein product
          View more View less
          Gene Aliases: Kcmb2; KCNMB2
          View more View less
          UniProt ID: (Human) Q9Y691, (Rat) Q811Q0
          View more View less
          Entrez Gene ID: (Human) 10242, (Rat) 294961
          View more View less
          Function(s)
          potassium channel activity protein binding ion channel inhibitor activity calcium-activated potassium channel activity potassium channel regulator activity voltage-gated ion channel
          Process(es)
          action potential detection of calcium ion ion transport potassium ion transport neuronal action potential regulation of vasoconstriction ion transmembrane transport potassium ion transmembrane transport chemical synaptic transmission
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-lnstr:80/100.66.74.145:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline