    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Kir2.2 (KCNJ12) Antibodies
          Alomone Labs, LTD
          Kir2.2 (KCNJ12) Polyclonal Antibody
          View all (10) Kir2.2 (KCNJ12) antibodies
          Tech Support
          Cite Kir2.2 (KCNJ12) Polyclonal Antibody
          Additional Information:
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (2)
          Kir2.2 (KCNJ12) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          Kir2.2 (KCNJ12) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          Kir2.2 (KCNJ12) Antibody (APC-042-25UL) in IHC (F)
          Expression ofKir2.2in ratcerebellum. Immunohistochemicalstaining of perfusion-fixed frozenratbrainsections with Anti-Kir2.2 (KCNJ12) Antibody &(8)239; (#APC-042), (1:300), followed by goat Anti-rabbit-AlexaFluor-488. A.Kir2.2immunoreactivity (green)appeared inBergmann glial fibers (right-pointing arrows) and outlines of molecular layer neurons (left-pointing arrows).B. Pre-incubation of the Antibody with Ki... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Kir2.2 (KCNJ12) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Kir2.2 (KCNJ12) Antibody in Western Blot (WB)
          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
          Kir2.2 (KCNJ12) Polyclonal Antibody
          Product Details
          APC-042-25UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Immunohistochemistry (Frozen) (IHC (F))
          1:300
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)EVATDRDGRSPQPEHDFDR,corresponding to amino acid residues 391-409 of rat Kir2.2, Intracellular, C-terminal domain
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.39 mg/mL
          Amount
          9.75 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          Inward rectifying potassium channel that is activated by phosphatidylinositol 4,5-bisphosphate and that probably participates in controlling the resting membrane potential in electrically excitable cells. Probably participates in establishing action potential waveform and excitability of neuronal and muscle tissues. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium.
          Bioinformatics
          Protein Aliases: inward rectifier K(+) channel Kir2.2; IRK-2; IRK2; potassium channel, inwardly rectifying subfamily J member 12; potassium channel, inwardly rectifying subfamily J, member 12; potassium voltage-gated channel subfamily J member 12; RB-IRK2; unnamed protein product
          View more View less
          Gene Aliases: hIRK; hIRK1; hkir2.2x; IRK-2; IRK2; kcnj12x; KCNJN1; Kir2.1; Kir2.2; MB-IRK2
          View more View less
          UniProt ID: (Human) Q14500, (Mouse) P52187
          View more View less
          Entrez Gene ID: (Human) 3768, (Mouse) 16515, (Rat) 117052
          View more View less
          Function(s)
          inward rectifier potassium channel activity protein binding PDZ domain binding
          Process(es)
          potassium ion transport protein homotetramerization potassium ion import across plasma membrane muscle contraction regulation of heart contraction
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-74d7b4b4d9-zrl6d:80/100.66.77.38:80.
          git-commit: 7aafa54bd143a040505a257d2a3ebda4095a617f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.56.0-Offline