    • See all product categories
  • Applications
    • Bioprocessing
  • Services
    • 360° CDMO and CRO Solutions
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • KCNH4 Antibodies
          Alomone Labs, LTD
          KV12.3 (KCNH4) Polyclonal Antibody
          View all (3) KCNH4 antibodies
          Tech Support
          Cite KV12.3 (KCNH4) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (2)
          KV12.3 (KCNH4) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          KV12.3 (KCNH4) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          FIGURE: 1 / 2
          KV12.3 (KCNH4) Antibody (APC-116-50UL) in IHC (P)
          Expression of KV12.3 (Elk3) in rat testes - Immunohistochemical staining of rat testes paraffin embedded section using Anti-KV12.3 (KCNH4) Antibody (#APC-116), (1:100).KV12.3 (pink staining) is expressed in the columnar epithelium of the epididymis. Hematoxilin is used as the counterstain. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          KV12.3 (KCNH4) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          KV12.3 (KCNH4) Antibody in Western Blot (WB)
          KV12.3 (KCNH4) Polyclonal Antibody
          Product Details
          APC-116-50UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Immunohistochemistry (Paraffin) (IHC (P))
          1:100
          -
          Product Specifications
          Species Reactivity
          Human, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          CRLNQEISRLNQEVS, corresponding to amino acids 881-895 of rat KCNH4, Intracellular, C-terminal domain
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          40 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit. The gene is brain-specific, and located in the neocortex and the striatum. It may be involved in cellular excitability of restricted neurons in the central nervous system.
          Bioinformatics
          Protein Aliases: BEC2; Brain-specific eag-like channel 2; ELK channel 1; ether-a-go-go K(+) channel family member; Ether-a-go-go-like potassium channel 1; potassium channel, voltage gated eag related subfamily H, member 4; Potassium voltage-gated channel subfamily H member 4; potassium voltage-gated channel, subfamily H (eag-related), member 4; rElk1; Voltage-gated delayed rectifier potassium channel KCNH4; Voltage-gated potassium channel subunit Kv12.3
          View more View less
          Gene Aliases: BEC2; ELK1; Elk3; KCNH4; Kv12.3
          View more View less
          UniProt ID: (Human) Q9UQ05, (Rat) Q9R1T9
          View more View less
          Entrez Gene ID: (Human) 23415, (Rat) 114032
          View more View less
          Function(s)
          ion channel activity voltage-gated potassium channel activity potassium channel activity phosphorelay sensor kinase activity ion channel
          Process(es)
          ion transport potassium ion transport ion transmembrane transport regulation of membrane potential transmembrane transport potassium ion transmembrane transport phosphorelay signal transduction system signal transduction by protein phosphorylation regulation of ion transmembrane transport
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Privacy Information Center
          • Legal Notice
          Germany flag icon
          Germany
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline