    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • CD69 Antibodies
          Proteintech
          CD69 Monoclonal Antibody (H1.2F3), APC
          View all (80) CD69 antibodies
          Tech Support
          Cite CD69 Monoclonal Antibody (H1.2F3), APC
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (1)
          CD69 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          CD69 Antibody in Flow Cytometry (Flow)
          Group 53 Created with Sketch.
          FIGURE: 1 / 1
          CD69 Antibody (APC-65105-100UG) in Flow
          1X10^6 PMA and ionomycin stimulated C57BL/6 Mouse splenocytes (6 hrs) were surface stained with 0.50 µg APC Anti-Mouse CD69 (Product # APC-65105-100UG), Clone: H1.2F3) (red) or 0.50 µg isotype control antibody (blue). Cells were not fixed. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          CD69 Antibody in Flow Cytometry (Flow)
          CD69 Monoclonal Antibody (H1.2F3), APC
          Product Details
          APC-65105-100UG
          Applications
          Tested Dilution
          Publications
          Flow Cytometry (Flow)
          Assay-dependent
          -
          Product Specifications
          Species Reactivity
          Mouse
          Host/Isotype
          Hamster / IgG
          Class
          Monoclonal
          Type
          Antibody
          Clone
          H1.2F3
          Conjugate
          APC APC APC
          • Unconjugated
          • FITC
          • PE 
          • Request custom conjugation
          Excitation/Emission Max
          651/660 nm View spectra spectra
          Form
          Liquid
          Concentration
          0.2 mg/mL
          Amount
          100 µg
          Purification
          Affinity chromatography
          Storage buffer
          PBS, pH 7.2, with 0.5% BSA
          Contains
          0.1% sodium azide
          Storage conditions
          4°C, store in dark
          Shipping conditions
          Wet ice
          Target Information
          CD69 (AIM, Active Inducer Molecule) is a gp28/34 disulfide bonded homodimer with a molecular weight of 60 kDa under non-reducing conditions. CD69 contains one or two N linked oligosacaride and the molecule is present on activated platelets. In normal peripheral blood a variable percentage of cells express the CD69 antigen, and it is involved in lymphocyte signal transduction. Expression CD69 is induced upon activation of T lymphocytes, and may play a role in proliferation. Furthermore, the protein may act to transmit signals in natural killer cells and platelets. Alternative splicing results in multiple transcript variants of CD69. Diseases associated with CD69 dysfunction include coccidiodomycosis and asthma.
          How to use the Panel Builder
          Bioinformatics
          Protein Aliases: CD69; Early activation antigen CD69; Very Early Activation Antigen
          View more View less
          Gene Aliases: 5830438K24Rik; AI452015; AIM; Cd69; VEA
          View more View less
          UniProt ID: (Mouse) P37217
          View more View less
          Entrez Gene ID: (Mouse) 12515
          View more View less
          Function(s)
          calcium ion binding carbohydrate binding
          Argentina flag icon
          Argentina
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-zsnhf:80/100.66.75.14:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline