Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Protein APC Antibodies

          Abnova

          APC Polyclonal Antibody, MaxPab™

          View all (40) Protein APC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite APC Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          APC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          APC Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          APC Antibody (H00000324-D01P) in WB

          Western Blot analysis of APC expression in transfected 293T cell line by APC MaxPab polyclonal antibody. Lane 1: APC transfected lysate(312.73 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          APC Antibody in Western Blot (WB)
          APC Polyclonal Antibody, MaxPab™

          Product Details

          H00000324-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Proximity Ligation Assay (PLA) (PLA)

          1:50-1:2,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          APC (AAI11592.1, 1 a.a. approximately 2843 a.a) full-length human protein.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.93 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAAASYDQLL KQVEALKMEN SNLRQELEDN SNHLTKLETE ASNMKEVLKQ LQGSIEDEAM ASSGQIDLLE RLKELNLDSS NFPGVKLRSK MSLRSYGSRE GSVSSRSGEC SPVPMGSFPR RGFVNGSRES TGYLEELEKE RSLLLADLDK EEKEKDWYYA QLQNLTKRID SLPLTENFSL QTDMTRRQLE YEARQIRVAM EEQLGTCQDM EKRAQRRIAR IQQIEKDILR IRQLLQSQAT EAERSSQNKH ETGSHDAERQ NEGQGVGEIN MATSGNGQGS TTRMDHETAS VLSSSSTHSA PRRLTSHLGT KVEMVYSLLS MLGTHDKDDM SRTLLAMSSS QDSCISMRQS GCLPLLIQLL HGNDKDSVLL GNSRGSKEAR ARASAALHNI IHSQPDDKRG RREIRVLHLL EQIRAYCETC WEWQEAHEPG MDQDKNPMPA PVEHQICPAV CVLMKLSFDE EHRHAMNELG GLQAIAELLQ VDCEMYGLTN DHYSITLRRY AGMALTNLTF GDVANKATLC SMKGCMRALV AQLKSESEDL QQVIASVLRN LSWRADVNSK KTLREVGSVK ALMECALEVK KESTLKSVLS ALWNLSAHCT ENKADICAVD GALAFLVGTL TYRSQTNTLA IIESGGGILR NVSSLIATNE DHRQILRENN CLQTLLQHLK SHSLTIVSNA CGTLWNLSAR NPKDQEALWD MGAVSMLKNL IHSKHKMIAM GSAAALRNLM ANRPAKYKDA NIMSPGSSLP SLHVRKQKAL EAELDAQHLS ETFDNIDNLS PKASHRSKQR HKQSLYGDYV FDTNRHDDNR SDNFNTGNMT VLSPYLNTTV LPSSSSSRGS LDSSRSEKDR SLERERGIGL GNYHPATENP GTSSKRGLQI STTAAQIAKV MEEVSAIHTS QEDRSSGSTT ELHCVTDERN ALRRSSAAHT HSNTYNFTKS ENSNRTCSMP YAKLEYKRSS NDSLNSVSSS DGYGKRGQMK PSIESYSEDD ESKFCSYGQY PADLAHKIHS ANHMDDNDGE LDTPINYSLK YSDEQLNSGR QSPSQNERWA RPKHIIEDEI KQSEQRQSRN QSTTYPVYTE STDDKHLKFQ PHFGQQECVS PYRSRGANGS ETNRVGSNHG INQNVSQSLC QEDDYEDDKP TNYSERYSEE EQHEEEERPT NYSIKYNEEK RHVDQPIDYS LKYATDIPSS QKQSFSFSKS SSGQSSKTEH MSSSSENTST PSSNAKRQNQ LHPSSAQSRS GQPQKAATCK VSSINQETIQ TYCVEDTPIC FSRCSSLSSL SSAEDEIGCN QTTQEADSAN TLQIAEIKEK IGTRSAEDPV SEVPAVSQHP RTKSSRLQGS SLSSESARHK AVEFSSGAKS PSKSGAQTPK SPPEHYVQET PLMFSRCTSV SSLDSFESRS IASSVQSEPC SGMVSGIISP SDLPDSPGQT MPPSRSKTPP PPPQTAQTKR EVPKNKAPTA EKRESGPKQA AVNAAVQRVQ VLPDADTLLH FATESTPDGF SCSSSLSALS LDEPFIQKDV ELRIMPPVQE NDNGNETESE QPKESNENQE KEAEKTIDSE KDLLDDSDDD DIEILEECII SAMPTKSSRK AKKPAQTASK LPPPVARKPS QLPVYKLLPS QNRLQPQKHV SFTPGDDMPR VYCVEGTPIN FSTATSLSDL TIESPPNELA AGEGVRGGAQ SGEFEKRDTI PTEGRSTDEA QGGKTSSVTI PELDDNKAEE GDILAECINS AMPKGKSHKP FRVKKIMDQV QQASASSSAP NKNQLDGKKK KPTSPVKPIP QNTEYRTRVR KNADSKNNLN AERVFSDNKD SKKQNLKNNS KVFNDKLPNN EDRVRGSFAF DSPHHYTPIE GTPYCFSRND SLSSLDFDDD DVDLSREKAE LRKAKENKES EAKVTSHTEL TSNQQSANKT QAIAKQPINR GQPKPILQKQ STFPQSSKDI PDRGAATDEK LQNFAIENTP VCFSHNSSLS SLSDIDQENN NKENEPIKET EPPDSQGEPS KPQASGYAPK SFHVEDTPVC FSRNSSLSSL SIDSEDDLLQ ECISSAMPKK KKPSRLKGDN EKHSPRNMGG ILGEDLTLDL KDIQRPDSEH GLSPDSENFD WKAIQEGANS IVSSLHQAAA AACLSRQASS DSDSILSLKS GISLGSPFHL TPDQEEKPFT SNKGPRILKP GEKSTLETKK IESESKGIKG GKKVYKSLIT GKVRSNSEIS GQMKQPLQAN MPSISRGRTM IHIPGVRNSS SSTSPVSKKG PPLKTPASKS PSEGQTATTS PRGAKPSVKS ELSPVARQTS QIGGSSKAPS RSGSRDSTPS RPAQQPLSRP IQSPGRNSIS PGRNGISPPN KLSQLPRTSS PSTASTKSSG SGKMSYTSPG RQMSQQNLTK QTGLSKNASS IPRSESASKG LNQMNNGNGA NKKVELSRMS STKSSGSESD RSERPVLVRQ STFIKEAPSP TLRRKLEESA SFESLSPSSR PASPTRSQAQ TPVLSPSLPD MSLSTHSSVQ AGGWRKLPPN LSPTIEYNDG RPAKRHDIAR SHSESPSRLP INRSGTWKRE HSKHSSSLPR VSTWRRTGSS SSILSASSES SEKAKSEDEK HVNSISGTKQ SKENQVSAKG TWRKIKENEF SPTNSTSQTV SSGATNGAES KTLIYQMAPA VSKTEDVWVR IEDCPINNPR SGRSPTGNTP PVIDSVSEKA NPNIKDSKDN QAKQNVGNGS VPMRTVGLEN RLNSFIQVDA PDQKGTEIKP GQNNPVPVSE TNESSIVERT PFSSSSSSKH SSPSGTVAAR VTPFNYNPSP RKSSADSTSA RPSQIPTPVN NNTKKRDSKT DSTESSGTQS PKRHSGSYLV TSV

          Target Information

          This gene encodes a tumor suppressor protein that acts as an antagonist of the Wnt signaling pathway. It is also involved in other processes including cell migration and adhesion, transcriptional activation, and apoptosis. Defects in this gene cause familial adenomatous polyposis (FAP), an autosomal dominant pre-malignant disease that usually progresses to malignancy. Disease-associated mutations tend to be clustered in a small region designated the mutation cluster region (MCR) and result in a truncated protein product.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: adenomatosis polyposis coli tumor suppressor; adenomatous polyposis coli; adenomatous polyposis coli (APC); Adenomatous polyposis coli protein; APC; APC, WNT signaling pathway regulator; Deleted in polyposis 2.5; epididymis secretory sperm binding protein; FAP; FPC; Protein APC; protein phosphatase 1, regulatory subunit 46; Unknown; unnamed protein product; WNT signaling pathway regulator

          View more View less

          Gene Aliases: APC; BTPS2; DESMD; DP2; DP2.5; DP3; GS; PPP1R46

          View more View less

          UniProt ID: (Human) P25054

          View more View less

          Entrez Gene ID: (Human) 324

          View more View less

          Function(s)
          protein binding beta-catenin binding microtubule binding protein kinase regulator activity protein kinase binding ubiquitin protein ligase binding macromolecular complex binding gamma-catenin binding cadherin binding microtubule plus-end binding dynein complex binding
          Process(es)
          mitotic cytokinesis cell fate specification heart valve development endocardial cushion morphogenesis cellular response to DNA damage stimulus negative regulation of microtubule depolymerization mitotic spindle assembly checkpoint cell adhesion pattern specification process nervous system development negative regulation of cell proliferation insulin receptor signaling pathway Wnt signaling pathway cell migration negative regulation of Wnt signaling pathway positive regulation of cell migration positive regulation of pseudopodium assembly regulation of microtubule-based process positive regulation of apoptotic process proteasome-mediated ubiquitin-dependent protein catabolic process positive regulation of protein catabolic process negative regulation of cyclin-dependent protein serine/threonine kinase activity regulation of attachment of spindle microtubules to kinetochore regulation of microtubule-based movement protein-containing complex assembly bicellular tight junction assembly regulation of primary metabolic process negative regulation of canonical Wnt signaling pathway positive regulation of cold-induced thermogenesis negative regulation of cell cycle G1/S phase transition positive regulation of protein localization to centrosome negative regulation of G1/S transition of mitotic cell cycle
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          France flag icon
          France

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline