Proteintech
Applications | Tested Dilution | Publications |
---|---|---|
Western Blot (WB) |
1:20,000-1:100,000 | View 2 publications 2 publications |
Immunohistochemistry (Paraffin) (IHC (P)) |
1:50-1:1,000 | - |
Immunocytochemistry (ICC/IF) |
1:50-1:500 | View 2 publications 2 publications |
Flow Cytometry (Flow) |
0.2 µg/1x10^6 cells | - |
Immunoprecipitation (IP) |
0.5-4.0 µg | - |
Product Specifications | |
---|---|
Species Reactivity |
Human |
Published species |
Human |
Host/Isotype |
Mouse / IgG2a |
Class |
Monoclonal |
Type |
Antibody |
Clone |
1G12D11 |
Immunogen |
APOL1-Specific Fusion Protein Ag2016 (1-238 aa encoded by BC017331) |
Conjugate |
Unconjugated
|
Form |
Liquid |
Concentration |
1.57 mg/mL |
Purification |
Protein A |
Storage buffer |
PBS, pH 7.3, with 50% glycerol |
Contains |
0.02% sodium azide |
Storage conditions |
-20°C |
This antibody recognizes the endogenous ApoL1 of 39-45 kDa in blood lysate. This antibody is specific to APOL1.
Immunogen sequence: MEGAALLRV SVLCIWMSAL FLGVGVRAEE AGARVQQNVP SGTDTGDPQS KPLGDWAAGT MDPESSIFIE DAIKYFKEKV STQNLLLLLT DNEAWNGFVA AAELPRNEAD ELRKALDNLA RQMIMKDKNW HDKGQQYRNW FLKEFPRLKS ELEDNIRRLR ALADGVQKVH KGTTIANVVS GSLSISSGIL TLVGMGLAPF TEGGSLVLLE PGMELGITAA LTGITSSTMD YGKKWWTQA (1-238 aa encoded by BC017331 )
APOL1 is a secreted high density lipoprotein which binds to apolipoprotein A-I. Apolipoprotein A-I is a relatively abundant plasma protein and is the major apoprotein of HDL. It is involved in the formation of most cholesteryl esters in plasma and also promotes efflux of cholesterol from cells. This apolipoprotein L family member may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Apo-L; APOL- I; ApoL-I; Apolipoprotein L; apolipoprotein L 1; apolipoprotein L, 1; Apolipoprotein L-I; Apolipoprotein L1; OTTHUMP00000028705
Gene Aliases: APO-L; APOL; APOL-I; APOL1; FSGS4
UniProt ID: (Human) O14791
Entrez Gene ID: (Human) 8542
Molecular Function:
apolipoprotein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support