Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: NQYENAEKLY LRSIAIGKKL FGEGYSGLEY DYRGLIKLYN SIGNYEKVFE YHNVLSNWNR LRDRQYSVTD ALEDVSTSPQ STEEVVQSFL ISQNVEGPSC
The protein encoded by this gene interacts with microtubules and is functionally associated with beta-amyloid precursor protein transport and/or processing. The beta-amyloid precursor protein is a cell surface protein with signal-transducing properties, and it is thought to play a role in the pathogenesis of Alzheimer's disease. This gene has been found to be highly expressed in breast cancer. Multiple polyadenylation sites have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: amyloid beta precursor protein (cytoplasmic tail) binding protein 2; Amyloid beta precursor protein-binding protein 2; Amyloid protein-binding protein 2; APP-BP2; Protein interacting with APP tail 1
Gene Aliases: APP-BP2; APPBP2; HS.84084; KIAA0228; PAT1
UniProt ID: (Human) Q92624
Entrez Gene ID: (Human) 10513
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support