    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • PLXNA3 Antibodies
          Alomone Labs, LTD
          Plexin-A3 (extracellular) Polyclonal Antibody
          View all (5) PLXNA3 antibodies
          Tech Support
          Cite Plexin-A3 (extracellular) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          Plexin-A3 (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          Plexin-A3 (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          Plexin-A3 (extracellular) Antibody (APR-093-50UL) in IHC (F)
          Expression of Plexin-A3 in mouse parietal cortex - Immunohistochemical staining of perfusion-fixed frozen mouse brain sections with Anti-Plexin-A3 (extracellular) Antibody (#APR-093), (1:200), followed by goat- Anti-rabbit-AlexaFluor-488. PLXNA3 staining (green) appears in pyramidal neurons (arrows). Cell nuclei are stained with DAPI (blue). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          Plexin-A3 (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Plexin-A3 (extracellular) Antibody in Western Blot (WB)
          Plexin-A3 (extracellular) Antibody in Western Blot (WB)
          Plexin-A3 (extracellular) Antibody in Flow Cytometry (Flow)
          Plexin-A3 (extracellular) Polyclonal Antibody
          Product Details
          APR-093-50UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Immunohistochemistry (Frozen) (IHC (F))
          1:200
          -
          Flow Cytometry (Flow)
          2.5 µg
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)KVRVDGSQDAQLYE, corresponding to amino acid residues 440-453 of mouse PLXNA3, Extracellular, N-terminus
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          40 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          Coreceptor for SEMA3A and SEMA3F. Necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. Plays a role in axon guidance in the developing nervous system. Regulates the migration of sympathetic neurons, but not of neural crest precursors. Required for normal dendrite spine morphology in pyramidal neurons. May play a role in regulating semaphorin-mediated programmed cell death in the developing nervous system. Class 3 semaphorins bind to a complex composed of a neuropilin and a plexin. The plexin modulates the affinity of the complex for specific semaphorins, and its cytoplasmic domain is required for the activation of down-stream signaling events in the cytoplasm.
          Bioinformatics
          Protein Aliases: plexin 3; Plexin 4, SEX, homolog to the cMet/HGF receptors; Plexin-4; Plexin-A3; Semaphorin receptor SEX; Sex chromosome X transmembrane protein of HGF receptor family 3; SEXPLEXIN-A3
          View more View less
          Gene Aliases: 6.3; HSSEXGENE; mKIAA4078; PlexA3; Plxa3; PLXN3; PLXN4; PLXNA3; SEX; XAP-6
          View more View less
          UniProt ID: (Human) P51805, (Mouse) P70208, (Rat) D3ZPX4
          View more View less
          Entrez Gene ID: (Human) 55558, (Mouse) 18846, (Rat) 309280
          View more View less
          Function(s)
          protein binding semaphorin receptor activity signaling receptor activity
          Process(es)
          signal transduction axon guidance facial nerve structural organization trigeminal nerve structural organization hippocampus development branchiomotor neuron axon guidance pyramidal neuron development negative regulation of axon extension involved in axon guidance negative chemotaxis positive regulation of cytoskeleton organization semaphorin-plexin signaling pathway involved in axon guidance cell surface receptor signaling pathway synapse assembly motor neuron axon guidance telencephalon development olfactory nerve formation gonadotrophin-releasing hormone neuronal migration to the hypothalamus regulation of cell migration axon extension regulation of axon extension involved in axon guidance axon extension involved in axon guidance regulation of axonogenesis regulation of chemotaxis semaphorin-plexin signaling pathway neuron projection guidance neuron projection extension
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline