Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • ARC Antibodies

          Rockland

          ARC Polyclonal Antibody

          View all (20) ARC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ARC Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          ARC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          ARC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ARC Antibody (600-401-Y55) in IHC (P)

          Immunohistochemistry of ARC antibody. Tissue: Human skeletal muscle. Fixation: formalin fixed paraffin embedded. Antigen retrieval: not required. Primary antibody: ARC antibody at 5 µg/mL for 1 hr at RT. Secondary antibody: Peroxidase rabbit secondary antibody at 1:10,000 for 45 min at RT. Localization: ARC is cytoplasmic and occasionally nuclear. Staining: ARC as precipitated pink signal with hematoxylin p... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ARC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ARC Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          ARC Antibody in Western Blot (WB)
          ARC Polyclonal Antibody

          Product Details

          600-401-Y55

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          5 µg/mL
          -

          ELISA (ELISA)

          1:10,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Anti-ARC antibody was prepared from whole rabbit serum produced by repeated immunizations with a 18 amino acid peptide near the C-terminus of human ARC.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.2

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          ARC (also known as NOL3 - Nucleolar protein 3) has two isoforms. Isoform 1 is involved in RNA splicing. Isoform 2 functions as an apoptosis repressor that blocks multiple modes of cell death. ARC inhibits extrinsic apoptotic pathways two different ways. The first is by interacting with FAS and FADD upon FAS activation blocking death-inducing signaling complex (DISC) assembly. The second way is by interacting with CASP8 in a mitochondria localization-and phosphorylation-dependent manner, which in turn limits the amount of soluble CASP8 avaliable for DISC-mediated activation. ARC also inhibits intrinsic apoptotic pathways in response to a wide range of stresses through interactions with BAX. This results in BAX inactivation and prevents mitochondrial dysfunction and release of pro-apoptotic factors. Mutations affecting the gene can results in myoclonus, familiam cortical.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Apoptosis repressor with CARD; muscle specific protein; Muscle-enriched cytoplasmic protein; Myp; Nop 30; Nop30; Nucleolar protein 3; nucleolar protein 3 (apoptosis repressor with CARD domain); Nucleolar protein of 30 kDa; unnamed protein product

          View more View less

          Gene Aliases: ARC; FCM; MYOCL1; MYP; NOL3; NOP; NOP30

          View more View less

          UniProt ID: (Human) O60936

          View more View less

          Entrez Gene ID: (Human) 8996

          View more View less

          Function(s)
          RNA binding receptor binding death receptor binding calcium ion binding protein binding death effector domain binding identical protein binding cysteine-type endopeptidase inhibitor activity involved in apoptotic process metal ion binding caspase binding
          Process(es)
          response to hypoxia blood vessel remodeling response to ischemia mRNA splice site selection mRNA processing apoptotic process RNA splicing negative regulation of signal transduction regulation of gene expression cardiac muscle cell apoptotic process negative regulation of striated muscle cell apoptotic process negative regulation of cardiac muscle cell apoptotic process negative regulation of tumor necrosis factor-mediated signaling pathway negative regulation of muscle atrophy release of sequestered calcium ion into cytosol by sarcoplasmic reticulum response to injury involved in regulation of muscle adaptation regulation of apoptotic process negative regulation of apoptotic process protein oligomerization negative regulation of programmed necrotic cell death negative regulation of release of cytochrome c from mitochondria intrinsic apoptotic signaling pathway regulation of NIK/NF-kappaB signaling negative regulation of mitochondrial membrane permeability involved in apoptotic process negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway regulation of intracellular signal transduction negative regulation of hypoxia-induced intrinsic apoptotic signaling pathway negative regulation of extrinsic apoptotic signaling pathway negative regulation of intrinsic apoptotic signaling pathway
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-5btrv:80/100.66.77.21:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline