Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • PARG1 Antibodies

          Abnova

          ARHGAP29 Polyclonal Antibody, MaxPab™

          View all (5) PARG1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ARHGAP29 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ARHGAP29 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ARHGAP29 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ARHGAP29 Antibody (H00009411-B01P) in WB

          Western Blot analysis of ARHGAP29 expression in transfected 293T cell line by ARHGAP29 MaxPab polyclonal antibody. Lane 1: ARHGAP29 transfected lysate(138.71 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ARHGAP29 Antibody in Western Blot (WB)

          Product Details

          H00009411-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ARHGAP29 (AAH93767.1, 1 a.a. approximately 1261 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MIAHKQKKTK KKRAWASGQL STDITTSEMG LKSLSSNSIF DPDYIKELVN DIRKFSHMLL YLKEAIFSDC FKEVIHIRLE ELLRVLKSIM NKHQNLNSVD LQNAAEMLTA KVKAVNFTEV NEENKNDLFQ EVFSSIETLA FTFGNILTNF LMGDVGNDSL LRLPVSRETK SFENVSVESV DSSSEKGNFS PLELDNVLLK NTDSIELALS YAKTWSKYTK NIVSWVEKKL NLELESTRNM VKLAEATRTN IGIQEFMPLQ SLFTNALLND IESSHLLQQT IAALQANKFV QPLLGRKNEM EKQRKEIKEL WKQEQNKMLE AENALKKAKL LCMQRQDEYE KAKSSMFRAE EEHLSSSGGL AKNLNKQLEK KRRLEEEALQ KVEEANELYK VCVTNVEERR NDLENTKREI LAQLRTLVFQ CDLTLKAVTV NLFHMQHLQA ASLADSLQSL CDSAKLYDPG QEYSEFVKAT NSTEEEKVDG NVNKHLNSSQ PSGFGPANSL EDVVRLPDSS NKIEEDRCSN SADITGPSFI RSWTFGMFSD SESTGGSSES RSLDSESISP GDFHRKLPRT PSSGTMSSAD DLDEREPPSP SETGPNSLGT FKKTLMSKAA LTHKFRKLRS PTKCRDCEGI VVFQGVECEE CLLVCHRKCL ENLVIICGHQ KLPGKIHLFG AEFTQVAKKE PDGIPFILKI CASEIENRAL CLQGIYRVCG NKIKTEKLCQ ALENGMHLVD ISEFSSHDIC DVLKLYLRQL PEPFILFRLY KEFIDLAKEI QHVNEEQETK KNSLEDKKWP NMCIEINRIL LKSKDLLRQL PASNFNSLHF LIVHLKRVVD HAEENKMNSK NLGVIFGPSL IRPRPTTAPI TISSLAEYSN QARLVEFLIT YSQKIFDGSL QPQDVMCSIG VVDQGCFPKP LLSPEERDIE RSMKSLFFSS KEDIHTSESE SKIFERATSF EESERKQNAL GKCDACLSDK AQLLLDQEAE SASQKIEDGK TPKPLSLKSD RSTNNVERHT PRTKIRPVSL PVDRLLLASP PNERNGRNMG NVNLDKFCKN PAFEGVNRKD AATTVCSKFN GFDQQTLQKI QDKQYEQNSL TAKTTMIMPS ALQEKGVTTS LQISGDHSIN ATQPSKPYAE PVRSVREASE RRSSDSYPLA PVRAPRTLQP QHWTTFYKPH APIISIRGNE EKPASPSAAV PPGTDHDPHG LVVKSMPDPD KASACPGQAT GQPKEDSEEL GLPDVNPMCQ RPRLKRMQQF EDLEDEIPQF V

          Target Information

          Rap1 is a small GTPase that, through effectors, regulates Rho GTPase signaling. These effectors- Rasip1, Radil, and the protein encoded by this gene- translocate to the cell membrane, where they form a multiprotein complex. This complex is necessary for Rap1-induced inhibition of Rho signaling. Defects in this gene may be a cause of nonsyndromic cleft lip with or without cleft palate.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: PARG; PTPL1-associated RhoGAP 1 (PARG1); PTPL1-associated RhoGAP protein 1; Rho GTPase-activating protein 29; Rho-type GTPase-activating protein 29; RP11-255E17.1

          View more View less

          Gene Aliases: ARHGAP29; PARG1

          View more View less

          UniProt ID: (Human) Q52LW3

          View more View less

          Entrez Gene ID: (Human) 9411

          View more View less

          Function(s)
          GTPase activator activity protein binding zinc ion binding PDZ domain binding metal ion binding
          Process(es)
          signal transduction Rho protein signal transduction regulation of small GTPase mediated signal transduction negative regulation of small GTPase mediated signal transduction
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0