Search
Search
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody is predicted to react with bovine based on sequence homology.
This antibody is tested in Peptide ELISA: antibody detection limit dilution 2,000.
ASNA1 is the human homolog of the bacterial arsA gene. In E. coli, ArsA ATPase is the catalytic component of a multisubunit oxyanion pump that is responsible for resistance to arsenicals and antimonials.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: arsA arsenite transporter, ATP-binding, homolog 1; arsenical pump-driving ATPase; arsenite-stimulated ATPase; arsenite-translocating ATPase; ATPase ASNA1; golgi to ER traffic 3 homolog; MGC3821; similar to bacterial and mouse ArsA; transmembrane domain recognition complex 40 kDa ATPase subunit; unnamed protein product
Gene Aliases: 1810048H22Rik; ArsA; ARSA-I; ARSA1; ASNA-I; ASNA1; CMD2H; TRC40
UniProt ID: (Human) O43681, (Rat) G3V9T7, (Mouse) O54984
Entrez Gene ID: (Human) 439, (Rat) 288919, (Mouse) 56495
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support