Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: LMTDRRIGC LLSGGLDSSL VAATLLKQLK EAQVQYPLQT FAIGMEDSPD LLAARKVADH IGSEHYEVLF NSEEGIQALD EVIFSLETYD ITTVRASVGM YLISKYIRKN TDSVVIFSGE GSDELTQGYI YFHKAPSPEK AEEESERLLR ELYLFDVLRA DRTTAAHGLE LRVPFLDHRF SSYYLSLPPE MRIPKNGIEK HLLRETFEDS NLIPKEILWR PKEAFSDGIT SVKNSWFKIL QEYVEHQVDD AMMANAAQKF PFNTPKTKEG YYYRQVFERH YPGRADWLSH YWMPKWINAT DPSARTLTHY KSAVKA (247-561 aa encoded by BC014621)
ASNS (asparagine synthetase [glutamine-hydrolyzing]) is involved in the subpathway that synthesizes f L-asparagine from L-aspartate. Mutations affecting the gene can result in asparagine synthetase deficiency (ASNSD).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: asparagine synthetase; glutamine-dependent asparagine synthetase; TS11 cell cycle control protein; unnamed protein product
Gene Aliases: ASNSD; TS11
UniProt ID: (Human) P08243
Entrez Gene ID: (Human) 440
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support