    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • GPR91 Antibodies
          Alomone Labs, LTD
          GPR91 (SUCNR1) (extracellular) Polyclonal Antibody
          View all (12) GPR91 antibodies
          Tech Support
          Cite GPR91 (SUCNR1) (extracellular) Polyclonal Antibody
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (3)
          GPR91 (SUCNR1) (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          GPR91 (SUCNR1) (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Group 53 Created with Sketch.
          FIGURE: 1 / 3
          GPR91 (SUCNR1) (extracellular) Antibody (ASR-090-50UL) in IHC (F)
          Expression of GPR91 in mouse hippocampus. Immunohistochemical staining of perfusion-fixed frozen mouse brain sections with Anti-GPR91 (SUCNR1) (extracellular) Antibody (#ASR-090), (1:200), followed by goat Anti-rabbit-AlexaFluor-488. A. GPR91 immunoreactivity (green) appears in neuronal profiles in the pyramidal layer (P, vertical arrow) and the stratum oriens (SO, horizontal arrow). B. Pre-incubation of th... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          GPR91 (SUCNR1) (extracellular) Antibody in Immunohistochemistry (Frozen) (IHC (F))
          GPR91 (SUCNR1) (extracellular) Antibody in Western Blot (WB)
          GPR91 (SUCNR1) (extracellular) Antibody in Flow Cytometry (Flow)
          GPR91 (SUCNR1) (extracellular) Polyclonal Antibody
          Product Details
          ASR-090-50UL
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:200
          -
          Immunohistochemistry (Frozen) (IHC (F))
          1:200
          -
          Flow Cytometry (Flow)
          2.5 µg/0.5x10^6 cells
          -
          Product Specifications
          Species Reactivity
          Human, Mouse, Rat
          Host/Isotype
          Rabbit / IgG
          Class
          Polyclonal
          Type
          Antibody
          Immunogen
          (C)RNVRIASRLDSWPQG, corresponding to amino acid residues 248-262 of rat GPR91/Succinate Receptor 1, 3rd extracellular loop
          3D Epitope / Immunogen
          Conjugate
          Unconjugated Unconjugated Unconjugated
          Form
          Lyophilized
          Concentration
          0.8 mg/mL
          Amount
          40 µg
          Purification
          Antigen affinity chromatography
          Storage buffer
          PBS, pH 7.4, with 1% BSA
          Contains
          0.05% sodium azide
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Product Specific Information
          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).
          Target Information
          This polyclonal antibody was raised in Rabbit against GPR91. This antibody is unconjugated. This antibody is useful for testing in the following assays: IHC-P. The specificity of this antibody has been described as follows: Human GPR91. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
          Bioinformatics
          Protein Aliases: G protein-coupled receptor 91; G-protein coupled receptor 91; G-protein-coupled receptor 91; p2u2; P2Y purinoceptor 1; P2Y purinoceptor 1-like; Succinate receptor; Succinate receptor 1; SUCR1; unnamed protein product
          View more View less
          Gene Aliases: GPR91; SUCNR1
          View more View less
          UniProt ID: (Human) Q9BXA5, (Rat) Q6IYF9, (Mouse) Q99MT6
          View more View less
          Entrez Gene ID: (Human) 56670, (Rat) 408199, (Mouse) 84112
          View more View less
          Function(s)
          G-protein coupled receptor activity protein binding signaling receptor activity signal transducer activity G-protein coupled receptor
          Process(es)
          renin secretion into blood stream macrophage activation involved in immune response signal transduction G-protein coupled receptor signaling pathway glucose homeostasis positive regulation of inflammatory response positive regulation of chemotaxis response to calcium ion regulation of angiotensin metabolic process energy homeostasis
          • News News
          • Responsibility Responsibility
          México flag icon
          México
          Your items have has been added!
          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline