Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • ASXL2 Antibodies

          Abnova

          ASXL2 Polyclonal Antibody, MaxPab™

          View all (9) ASXL2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite ASXL2 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          ASXL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          ASXL2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          ASXL2 Antibody (H00055252-B01P) in WB

          Western Blot analysis of ASXL2 expression in transfected 293T cell line by ASXL2 MaxPab polyclonal antibody. Lane 1: ASXL2 transfected lysate(100.98 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          ASXL2 Antibody in Western Blot (WB)

          Product Details

          H00055252-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          ASXL2 (AAH42999, 1 a.a. approximately 918 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.38 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MKRTKCADID VETPDSILVN TNLRALINKH TFSVLPGDCQ QRLLLLLPEV DRQVGPDGLM KLNGSALNNE FFTSAAQGWK ERLSEGEFTP EMQVRIRQEI EKEKKVEPWK EQFFESYYGQ SSGLSLEDSK KLTASPSDPK VKKTPAEQPK SMPVSEASLI RIVPVVSQSE CKEEALQMSS PGRKEECESQ GEVQPNFSTS SEPLLSSALN THELSSILPI KCPKDEDLLE QKPVTSAEQE SEKNHLTTAS NYNKSESQES LVTSPSKPKS PGVEKPIVKP TAGAGPQETN MKEPLATLVD QSPESLKRKS SLTQEEAPVS WEKRPRVTEN RQHQQPFQVS PQPFLNRGDR IQVRKVPPLK IPVSRISPMP FHPSQVSPRA RFPVSITSPN RTGARTLADI KAKAQLVKAQ RAAAAAAAAA AAAASVGGTI PGPGPGGGQG PGEGGEGQTA RGGSPGSDRV SETGKGPTLE LAGTGSRGGT RELLPCGPET QPQSETKTTP SQAQPHSVSG AQLQQTPPVP PTPAVSGACT SVPSPVHIEK LDNEKLNPTR ATATVASVSH PQGPSSCRQE KAPSPTGFQL EDISTSQRFM LGFAGRRTSK PAMAGHYLLN ISTYGRGSES FRRTHSVNPE DRFCLSSPTE ALKMGYTDCK NATGESSSSK EDDTDEESTG DEQESVTVKE EPQVSQSAGK GDTSSGPHSR ETLSTSDCLA SKNVKAEIPL NEQTTLSKEN YLFTRGQTFD EKTLARDLIQ AAQKQMAHAV RGKAIRSSPE LFSSTVLPLP ADSPTHQPLL LPPLQTPKLY GSPTQIGPSY RGMINVSTSS DMDHNSAVPG SQVSSNVGDV MSFSVTVTTI PASQAMNPSS HGQTIPVQAF SEENSIEGTP SKCYCRLKAM IMCKGCGAFC HDDCIGPSKL CVSCLVVR

          Target Information

          Putative Polycomb group (PcG) protein. PcG proteins act by forming multiprotein complexes, which are required to maintain the transcriptionally repressive state of homeotic genes throughout development. PcG proteins are not required to initiate repression, but to maintain it during later stages of development. They probably act via methylation of histones, rendering chromatin heritably changed in its expressibility. Involved in transcriptional regulation mediated by ligand-bound nuclear hormone receptors, such as peroxisome proliferator-activated receptor gamma (PPARG). Acts as coactivator for PPARG and enhances its adipocyte differentiation-inducing activity; the function seems to involve differential recruitment of acetylated and methylated histone H3.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: additional sex combs like 2, transcriptional regulator; Additional sex combs-like protein 2; polycomb group protein ASXH2; Putative Polycomb group protein ASXL2; unnamed protein product

          View more View less

          Gene Aliases: ASXH2; ASXL2; KIAA1685; SHAPNS

          View more View less

          UniProt ID: (Human) Q76L83

          View more View less

          Entrez Gene ID: (Human) 55252

          View more View less

          Function(s)
          DNA binding chromatin binding protein binding zinc ion binding peroxisome proliferator activated receptor binding metal ion binding
          Process(es)
          regulation of transcription, DNA-templated organ morphogenesis positive regulation of peroxisome proliferator activated receptor signaling pathway positive regulation of fat cell differentiation positive regulation of transcription from RNA polymerase II promoter
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-b2k9d:80/100.66.79.173:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline