Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • ADORA2A Antibodies

          Alomone Labs, LTD

          Adenosine A2A Receptor Polyclonal Antibody

          View all (18) ADORA2A antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Adenosine A2A Receptor Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          Adenosine A2A Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Adenosine A2A Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Adenosine A2A Receptor Antibody (AAR-002-200UL) in IHC (P)

          Expression of AdenosineA2AReceptorin rat lung - Immunohistochemical staining of paraffin embedded rat lung sections using Anti-AdenosineA2AReceptor Antibody (#AAR-002), (1:50). A2aR is expressed in the respiratory epithelium of the bronchiole (Br). Note that smooth muscle and endothelium in blood vessels are negative. Hematoxilin is used as the counterstain. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Adenosine A2A Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Adenosine A2A Receptor Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Adenosine A2A Receptor Antibody in Western Blot (WB)
          Adenosine A2A Receptor Polyclonal Antibody

          Product Details

          AAR-002-200UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:50
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          (C)RQLKQMESQPLPGER, corresponding to amino acid residues 201-215 of mouse adenosine A2Areceptor, 3rd intracellular loop
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          0.75 mg/mL

          Amount

          150 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 1% BSA

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitution: 25 µL, 50 µL or 0.2 mL double distilled water (DDW), depending on the sample size. The antibody ships as a lyophilized powder at room temperature. Upon arrival, it should be stored at -20C. The reconstituted solution can be stored at 4C for up to 1 week. For longer periods, small aliquots should be stored at -20C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min).

          Target Information

          ADORA2A is a protein which is one of several receptor subtypes for adenosine. The activity of the encoded protein, a G-protein coupled receptor family member, is mediated by G proteins which activate adenylyl cyclase. This protein is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: A2a receptor; Adenosine A2a-receptor; Adenosine receptor A2a; adenosine receptor subtype A2a; ADORA2A; AR; hA2aR; unnamed protein product

          View more View less

          Gene Aliases: A2AAR; A2aR; AA2AR; ADENO; ADORA2; ADORA2A; Adora2l1; RDC8

          View more View less

          UniProt ID: (Human) P29274, (Mouse) Q60613, (Rat) P30543

          View more View less

          Entrez Gene ID: (Human) 135, (Mouse) 11540, (Rat) 25369

          View more View less

          Function(s)
          G-protein coupled adenosine receptor activity G-protein coupled receptor activity protein binding calmodulin binding lipid binding enzyme binding type 5 metabotropic glutamate receptor binding identical protein binding macromolecular complex binding alpha-actinin binding signal transducer activity protein heterodimerization activity G-protein coupled receptor
          Process(es)
          synaptic transmission, dopaminergic adenosine receptor signaling pathway response to amphetamine regulation of transcription, DNA-templated phagocytosis apoptotic process inflammatory response cellular defense response signal transduction G-protein coupled receptor signaling pathway adenylate cyclase-modulating G-protein coupled receptor signaling pathway adenylate cyclase-activating G-protein coupled receptor signaling pathway phospholipase C-activating G-protein coupled receptor signaling pathway cell-cell signaling synaptic transmission, cholinergic central nervous system development blood coagulation sensory perception locomotory behavior blood circulation negative regulation of cell proliferation response to xenobiotic stimulus positive regulation of glutamate secretion positive regulation of acetylcholine secretion, neurotransmission regulation of norepinephrine secretion response to purine-containing compound response to caffeine positive regulation of synaptic transmission, GABAergic synaptic transmission, glutamatergic positive regulation of urine volume vasodilation eating behavior negative regulation of vascular permeability response to alkaloid negative regulation of neuron apoptotic process positive regulation of circadian sleep/wake cycle, sleep negative regulation of alpha-beta T cell activation astrocyte activation neuron projection morphogenesis positive regulation of protein secretion negative regulation of inflammatory response regulation of mitochondrial membrane potential membrane depolarization regulation of calcium ion transport positive regulation of synaptic transmission, glutamatergic excitatory postsynaptic potential inhibitory postsynaptic potential prepulse inhibition positive regulation of ERK1 and ERK2 cascade apoptotic signaling pathway presynaptic modulation of chemical synaptic transmission positive regulation of long-term synaptic potentiation positive regulation of apoptotic signaling pathway negative regulation of protein kinase activity protein kinase C-activating G-protein coupled receptor signaling pathway positive regulation of adenylate cyclase activity involved in G-protein coupled receptor signaling pathway positive regulation of cAMP biosynthetic process positive regulation of renal sodium excretion negative regulation of locomotion negative regulation of cysteine-type endopeptidase activity involved in apoptotic process regulation of synaptic plasticity response to drug
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-6b469b8bb7-lcddp:80/100.66.76.150:80.
          git-commit: a334af76dff23450325448aedefe62379591458a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.1-2026.04.16-1.0