Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
This antibody raised against 1-147aa of human AdipoR2 may cross-react with AdipoR1.
Immunogen sequence: MNEPTENRL GCSRTPEPDI RLRKGHQLDG TRRGDNDSHQ GDLEPILEAS VLSSHHKKSS EEHEYSDEAP QEDEGFMGMS PLLQAHHAME KMEEFVCKVW EGRWRVIPHD VLPDWLKDND FLLHGHRPPM PSFRACFKSI FRIHTETG (1-147 aa encoded by BC051858)
Adiponectin (also known as 30- kDa adipocyte complement-related protein; Acrp30) is a hormone secreted by adipocytes that acts as an antidiabetic and anti-atherogenic adipokine. Levels of adiponectin in the blood are decreased under conditions of obesity, insulin resistance and type 2 diabetes. Administration of adiponectin causes glucose-lowering effects and ameliorates insulin resistance in mice. Conversely, adiponectin-deficient mice exhibit insulin resistance and diabetes. This insulin-sensitizing effect of adiponectin seems to be mediated by an increase in fatty-acid oxidation through activation of AMP kinase and PPAR-. Here we report the cloning of complementary DNAs encoding adiponectin receptors 1 and 2 (AdipoR1 and AdipoR2) by expression cloning. AdipoR1 is abundantly expressed in skeletal muscle, whereas AdipoR2 is predominantly expressed in the liver. These two adiponectin receptors are predicted to contain seven transmembrane domains, but are structurally and functionally distinct from G-protein-coupled receptors. Expression of AdipoR1/R2 or suppression of AdipoR1/R2 expression by small-interfering RNA supports our conclusion that they serve as receptors for globular and full-length adiponectin, and that they mediate increased AMP kinase and PPAR- ligand activities, as well as fatty-acid oxidation and glucose uptake by adiponectin.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: progestin and adipoQ receptor family member 1; progestin and adipoQ receptor family member 2; progestin and adipoQ receptor family member I; progestin and adipoQ receptor family member II; receptor; unnamed protein product
Gene Aliases: 1110001I14Rik; 2810031L11Rik; ACDCR1; ACDCR2; ADCR2; CGI-45; CGI45; D6Ucla1e; PAQR1; PAQR2; TESBP1A
UniProt ID: (Human) Q96A54, (Human) Q86V24, (Mouse) Q91VH1, (Mouse) Q8BQS5
Entrez Gene ID: (Human) 51094, (Human) 79602, (Rat) 289036, (Mouse) 72674, (Mouse) 68465, (Rat) 312670
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support