Search
Search
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.
Immunogen sequence: MLLELTRRG QEPLSALLQF GVTYLEDYSA EYIIQQGGWG TVFSLESEEE EYPGITAEDS NDIYILPSDN SGQVSPPESP TVTTSWQSES LPVSLSASQS WHTESLPVSL GPESWQQIAM DPEEVKSLDS NGAGEKSENN SSNSDIVHVE KEEVPEGMEE AAVASVVLPA RELQEALPEA PAPLLPHITA TSLLGTREPD TEVITVEKSS PATSLFVELD EEEVKAATTE PTEVEEVVPA LEPTETLLSE KEINAREESL VEELSPASEK KPVPPSEGKS RLSPAGEMKP MPLSEGKSIL L (1-300 aa encoded by BC003032)
Apoptosis plays a major role in normal organism development, tissue homeostasis, and removal of damaged cells. Disruption of this process has been implicated in a variety of diseases such as cancer. Members of the Bcl-2 family are known to be critical regulators of this process. These proteins are characterized by the presence of several conserved motifs termed Bcl-2 homology (BH) domains. A novel, widely expressed member termed Bcl-rambo was recently identified. This protein is localized to mitochondria in mammalian cells and its overexpression induces apoptosis which could be blocked by co-expression of inhibitor of apoptosis proteins (IAPs) such as XIAP, cIAP1, and cIAP2. Bcl-rambo shows overall homology to the anti-apoptotic members containing BH motifs, but unlike Bcl-2, the C-terminal membrane anchor of Bcl-rambo is preceded by a unique 250 amino acid insertion. This region by itself can induce apoptosis more efficiently than the Bcl-2 homology regions, suggesting that Bcl-rambo may be important other pro-apoptotic pathways.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bcl2-L-13; BCL2-like 13 (apoptosis facilitator); MIL1; mitochondria located 1 homolog; protein Mil1; unnamed protein product
Gene Aliases: BCL-RAMBO; Bcl2-L-13; E430016C20Rik; Mil-1; MIL1
UniProt ID: (Human) Q9BXK5, (Mouse) P59017
Entrez Gene ID: (Human) 23786, (Rat) 312682, (Mouse) 94044
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn more
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support