Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          • Primary Antibodies ›
          • BCL-3 Antibodies

          Abnova

          BCL3 Polyclonal Antibody, MaxPab™

          View all (13) BCL-3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite BCL3 Polyclonal Antibody, MaxPab™

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (1)
          BCL3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          BCL3 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BCL3 Antibody (H00000602-B01P) in WB

          Western Blot analysis of BCL3 expression in transfected 293T cell line by BCL3 MaxPab polyclonal antibody. Lane 1: BCL3 transfected lysate(49.06 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BCL3 Antibody in Western Blot (WB)

          Product Details

          H00000602-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          BCL3 (NP_005169.1, 1 a.a. approximately 446 a.a) full-length human protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          50 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MDEGPVDLRT RPKAAGLPGA ALPLRKRPLR APSPEPAAPR GAAGLVVPLD PLRGGCDLPA VPGPPHGLAR PEALYYPGAL LPLYPTRAMG SPFPLVNLPT PLYPMMCPME HPLSADIAMA TRADEDGDTP LHIAVVQGNL PAVHRLVNLF QQGGRELDIY NNLRQTPLHL AVITTLPSVV RLLVTAGASP MALDRHGQTA AHLACEHRSP TCLRALLDSA APGTLDLEAR NYDGLTALHV AVNTECQETV QLLLERGADI DAVDIKSGRS PLIHAVENNS LSMVQLLLQH GANVNAQMYS GSSALHSASG RGLLPLVRTL VRSGADSSLK NCHNDTPLMV ARSRRVIDIL RGKATRPAST SQPDPSPDRS ANTSPESSSR LSSNGLLSAS PSSSPSQSPP RDPPGFPMAP PNFFLPSPSP PAFLPFAGVL RGPGRPVPPS PAPGGS

          Target Information

          B cell Lymphoma 3 is a member of the IkB subfamily of inhibitors. This subfamily is part of the NF-kB transcription factor protein family and it is suggested that a balance in the concentration of various NF-kB family members regulates apoptosis and survival of activated T cells. Bcl-3 has been shown to have versatile functions such as cytoplasmic activation of p50 homodimers, translocation to the nucleus, and modulation of the transcriptional machinery in the nucleus. Bcl-3 activity often relies on several nuclear interacting proteins such as Tip60, Jab1, Bard1, and Pirin.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: B cell CLL/lymphoma 3; B-cell leukemia/lymphoma 3; B-cell lymphoma 3 protein; B-cell lymphoma 3-encoded protein; BCL-3; Bcl-3 protein; chronic lymphatic leukemia protein; lymphoma 3-encoded protein (bcl-3); Proto-oncogene BCL3; unnamed protein product

          View more View less

          Gene Aliases: BCL3; BCL4; D19S37

          View more View less

          UniProt ID: (Human) P20749

          View more View less

          Entrez Gene ID: (Human) 602

          View more View less

          Function(s)
          transcription cofactor activity transcription coactivator activity transcription corepressor activity protein binding protein binding, bridging histone deacetylase binding DNA-binding transcription factor binding
          Process(es)
          negative regulation of cytokine production follicular dendritic cell differentiation marginal zone B cell differentiation humoral immune response mediated by circulating immunoglobulin germinal center formation protein import into nucleus apoptotic process response to stress cellular response to DNA damage stimulus I-kappaB kinase/NF-kappaB signaling response to virus response to UV-C positive regulation of gene expression antimicrobial humoral response extracellular matrix organization DNA damage response, signal transduction by p53 class mediator negative regulation of NF-kappaB transcription factor activity negative regulation of interleukin-8 production negative regulation of tumor necrosis factor production positive regulation of interferon-gamma production positive regulation of interleukin-10 production T-helper 1 type immune response defense response to bacterium intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator defense response to protozoan regulation of apoptotic process negative regulation of apoptotic process T-helper 2 cell differentiation positive regulation of translation negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter negative regulation of JAK-STAT cascade spleen development regulation of DNA binding response to other organism T cell apoptotic process negative regulation of T cell apoptotic process regulation of NIK/NF-kappaB signaling
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-8dsbr:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline