Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Bcr Antibodies

          Proteintech

          BCR Polyclonal Antibody

          2 References
          View all (30) Bcr antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite BCR Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (12)
          BCR Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          BCR Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BCR Antibody (22585-1-AP) in ICC/IF

          Immunofluorescent analysis of ( -20°C Ethanol ) fixed HeLa cells using 22585-1-AP (BCR antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BCR Antibody in Immunocytochemistry (ICC/IF)
          BCR Antibody in Immunocytochemistry (ICC/IF)
          BCR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BCR Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BCR Antibody in Western Blot (WB)
          BCR Antibody in Western Blot (WB)
          BCR Antibody in Western Blot (WB)
          BCR Antibody in Western Blot (WB)
          BCR Antibody in Western Blot (WB)
          BCR Antibody in Western Blot (WB)
          BCR Antibody in Flow Cytometry (Flow)
          BCR Antibody in Immunoprecipitation (IP)
          BCR Polyclonal Antibody

          Product Details

          22585-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 1 publication 1 publication

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:500
          -

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -
          Product Specifications

          Species Reactivity

          Human, Non-human primate

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          BCR Fusion Protein Ag18281 (1-350 aa encoded by BC066122)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.45 mg/mL

          Amount

          67.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MVDPVGFAE AWKAQFPDSE PPRMELRSVG DIEQELERCK ASIRRLEQEV NQERFRMIYL QTLLAKEKKS YDRQRWGFRR AAQAPDGASE PRASASRPQP APADGADPPP AEEPEARPDG EGSPGKARPG TARRPGAAAS GERDDRGPPA SVAALRSNFE RIRKGHGQPG ADAEKPFYVN VEFHHERGLV KVNDKEVSDR ISSLGSQAMQ MERKKSQHGA GSSVGDASRP PYRGRSSESS CGVDGDYEDA ELNPRFLKDN LIDANGGSRP PWPPLEYQPY QSIYVGGMME GEGKGPLLRS QSTSEQEKRL TWPRRSYSPR SFEDCGGGYT PDCSSNENLT SSEEDFSSGQ S (1-350 aa encoded by BC066122)

          Target Information

          A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ABL; bcr; BCR protein; BCR, RhoGEF and GTPase activating protein; bcr/abl; bcr/able fusion protein; BCR/FGFR1 chimera protein; breakpoint cluster region; Breakpoint cluster region protein; c-ABL; c-abl oncogene 1; EC 2.7.11.1; FGFR1/BCR chimera protein; JTK7; p150; protein kinase; Renal carcinoma antigen NY-REN-26; RP11-83J21.1; unnamed protein product; v-abl

          View more View less

          Gene Aliases: ALL; BCR; BCR1; CML; D22S11; D22S662; PHL

          View more View less

          UniProt ID: (Human) P11274

          View more View less

          Entrez Gene ID: (Human) 613

          View more View less

          Function(s)
          nucleotide binding protein serine/threonine kinase activity protein tyrosine kinase activity guanyl-nucleotide exchange factor activity GTPase activator activity protein binding ATP binding kinase activity transferase activity protein serine kinase activity
          Process(es)
          negative regulation of cellular extravasation renal system process protein phosphorylation phagocytosis signal transduction small GTPase mediated signal transduction brain development cell migration actin cytoskeleton organization keratinocyte differentiation response to lipopolysaccharide regulation of Rho protein signal transduction intracellular signal transduction inner ear morphogenesis regulation of vascular permeability neutrophil degranulation negative regulation of neutrophil degranulation focal adhesion assembly homeostasis of number of cells negative regulation of inflammatory response positive regulation of phagocytosis modulation of synaptic transmission neuromuscular process controlling balance regulation of small GTPase mediated signal transduction establishment of localization in cell regulation of cell cycle definitive hemopoiesis negative regulation of respiratory burst negative regulation of blood vessel remodeling intracellular protein transmembrane transport cellular response to lipopolysaccharide activation of GTPase activity macrophage migration negative regulation of macrophage migration negative regulation of reactive oxygen species metabolic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
          git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.48.0-Offline