Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          • Primary Antibodies ›
          • BMP4 Antibodies

          Bioss

          BMP4 Polyclonal Antibody

          View all (20) BMP4 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite BMP4 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          BMP4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          BMP4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BMP4 Antibody (BS-1374R) in IHC (P)

          Paraformaldehyde-fixed, paraffin embedded mouse pancreas; Antigen retrieval by boiling in sodium citrate buffer (pH6.0) for 15min; Block endogenous peroxidase by 3% hydrogen peroxide for 20 minutes; Blocking buffer (normal goat serum) at 37°C for 30min; Antibody incubation with BMP4 Polyclonal Antibody, Unconjugated (bs-1374R) at 1:400 overnight at 4°C, followed by a conjugated secondary for 20 minutes and ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BMP4 Antibody in Immunohistochemistry (Paraffin) (IHC (P))

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          BMP4 Polyclonal Antibody

          Product Details

          BS-1374R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:400
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Mouse, Sheep, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic peptide derived from rat BMP4, amino acids 251-350.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 1% BSA, 50% glycerol

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5' untranslated region of this gene has been described and three variants are described, all encoding an identical protein.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BMP; BMP-2B; BMP-4; BMP4; Bone morphogenetic protein; bone morphogenetic protein 2B; unnamed protein product

          View more View less

          Gene Aliases: Bmp-4; Bmp2b; Bmp2b-1; Bmp2b1; BOMPR4A

          View more View less

          UniProt ID: (Mouse) P21275

          View more View less

          Entrez Gene ID: (Sheep) 443502, (Mouse) 12159, (Rat) 25296

          View more View less

          Function(s)
          cytokine activity protein binding growth factor activity heparin binding co-receptor binding chemoattractant activity identical protein binding protein serine/threonine kinase activator activity BMP receptor binding growth factor
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter skeletal system development angiogenesis ovarian follicle development blood vessel development osteoblast differentiation eye development metanephros development ureteric bud development branching involved in ureteric bud morphogenesis mesoderm formation organ induction kidney development mesonephros development neural tube closure positive regulation of protein phosphorylation regulation of endothelial cell proliferation positive regulation of endothelial cell proliferation vasculature development endochondral ossification blood vessel endothelial cell proliferation involved in sprouting angiogenesis chondrocyte differentiation hematopoietic progenitor cell differentiation lymphoid progenitor cell differentiation heart morphogenesis renal system process heart induction secondary heart field specification outflow tract septum morphogenesis membranous septum morphogenesis muscular septum morphogenesis outflow tract morphogenesis aortic valve morphogenesis pulmonary valve morphogenesis endocardial cushion development epithelial to mesenchymal transition involved in endocardial cushion formation cardiac right ventricle morphogenesis apoptotic process involved in endocardial cushion morphogenesis type B pancreatic cell development germ cell development endoderm development mesoderm development mesodermal cell fate determination heart development positive regulation of cell proliferation negative regulation of cell proliferation post-embryonic development anterior/posterior axis specification specification of organ position regulation of cell fate commitment gene expression regulation of gene expression positive regulation of calcium ion transport into cytosol positive regulation of endothelial cell migration positive regulation of gene expression negative regulation of gene expression positive regulation of epithelial to mesenchymal transition positive regulation of neuron projection development cardiac muscle hypertrophy in response to stress telencephalon development dorsal/ventral neural tube patterning telencephalon regionalization pituitary gland development erythrocyte differentiation monocyte differentiation macrophage differentiation lung development embryonic limb morphogenesis positive regulation of cell migration positive regulation of bone mineralization BMP signaling pathway regulation of BMP signaling pathway positive regulation of BMP signaling pathway negative regulation of BMP signaling pathway forebrain development positive regulation of vascular endothelial growth factor receptor signaling pathway negative regulation of chondrocyte differentiation positive regulation of collagen biosynthetic process negative regulation of T cell differentiation in thymus negative regulation of immature T cell proliferation in thymus cellular response to oxidative stress embryonic hindlimb morphogenesis tendon cell differentiation deltoid tuberosity development specification of segmental identity, mandibular segment regulation of protein import into nucleus odontogenesis of dentin-containing tooth odontogenesis regulation of odontogenesis of dentin-containing tooth embryonic digit morphogenesis camera-type eye development positive regulation of apoptotic process positive regulation of programmed cell death regulation of MAPK cascade cell fate commitment regulation of cell differentiation positive regulation of cell differentiation positive regulation of endothelial cell differentiation positive regulation of epidermal cell differentiation negative regulation of myoblast differentiation positive regulation of neuron differentiation positive regulation of osteoblast differentiation positive regulation of ossification negative regulation of cell cycle negative regulation of mitotic nuclear division negative regulation of striated muscle tissue development negative regulation of transcription, DNA-templated positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter lung alveolus development mesodermal cell differentiation intermediate mesodermal cell differentiation camera-type eye morphogenesis embryonic morphogenesis anatomical structure formation involved in morphogenesis regulation of smooth muscle cell proliferation positive regulation of smooth muscle cell proliferation neuron fate commitment embryonic cranial skeleton morphogenesis embryonic skeletal system morphogenesis embryonic skeletal system development negative regulation of oligodendrocyte differentiation smooth muscle tissue development stem cell differentiation positive regulation of epithelial cell proliferation negative regulation of epithelial cell proliferation positive chemotaxis smooth muscle cell differentiation regulation of smooth muscle cell differentiation negative regulation of multicellular organismal process cardiac muscle cell differentiation positive regulation of cardiac muscle fiber development inner ear receptor cell differentiation lens induction in camera-type eye embryonic skeletal joint morphogenesis bone development cranial suture morphogenesis positive regulation of SMAD protein import into nucleus lung morphogenesis lung vasculature development epithelium development bronchus development trachea development trachea formation epithelial tube branching involved in lung morphogenesis branching involved in prostate gland morphogenesis bud elongation involved in lung branching epithelial cell proliferation involved in lung morphogenesis bud dilation involved in lung branching prostate gland morphogenesis mammary gland formation epithelial-mesenchymal cell signaling negative regulation of prostatic bud formation regulation of branching involved in prostate gland morphogenesis regulation of morphogenesis of a branching structure coronary vasculature development regulation of cartilage development positive regulation of cartilage development positive regulation of branching involved in lung morphogenesis pharyngeal arch artery morphogenesis negative regulation of thymocyte apoptotic process positive regulation of hepatocyte differentiation regulation of ERK1 and ERK2 cascade positive regulation of ERK1 and ERK2 cascade cellular response to growth factor stimulus cellular response to dexamethasone stimulus cellular response to BMP stimulus glomerular capillary formation negative regulation of glomerular mesangial cell proliferation nephric duct formation ureter morphogenesis negative regulation of mesenchymal cell proliferation involved in ureter development regulation of branching involved in ureteric bud morphogenesis positive regulation of branching involved in ureteric bud morphogenesis negative regulation of branching involved in ureteric bud morphogenesis negative regulation of glomerulus development sensory organ morphogenesis positive regulation of protein localization to nucleus positive regulation of p38MAPK cascade positive regulation of odontoblast differentiation positive regulation of store-operated calcium channel activity positive regulation of mesenchymal stem cell proliferation negative regulation of pri-miRNA transcription from RNA polymerase II promoter positive regulation of pri-miRNA transcription from RNA polymerase II promoter cardiac jelly development positive regulation of cardiac neural crest cell migration involved in outflow tract morphogenesis positive regulation of morphogenesis of an epithelium negative regulation of metanephric S-shaped body morphogenesis negative regulation of metanephric comma-shaped body morphogenesis positive regulation of endothelial cell apoptotic process positive regulation of primary miRNA processing negative regulation of extrinsic apoptotic signaling pathway response to mechanical stimulus positive regulation of epithelial cell differentiation response to estradiol response to retinoic acid response to testosterone tongue morphogenesis response to glucocorticoid anatomical structure regression retina development in camera-type eye lung lobe development cellular response to mechanical stimulus response to nitric oxide cellular response to thyroid hormone stimulus regulation of miRNA transcription
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-green-74676f44b6-dg9d8:80/100.66.76.64:80.
          git-commit: db542085ed6beb0c3f0dd774c2548ab49b9f1b6a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026.10.42-1.0