Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • BMPR2 Antibodies

          Proteintech

          BMPR2 Polyclonal Antibody

          7 References
          View all (14) BMPR2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite BMPR2 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          BMPR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          BMPR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 6

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          BMPR2 Antibody (19087-1-AP) in IHC (P)

          Immunohistochemical analysis of paraffin-embedded Human heart tissue slide using (Product # 19087-1-AP) BMPR2 antibody at dilution of 1:200 (under 40x lens). Heat mediated antigen retrieval with Tris-EDTA buffer (pH 9.0). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          BMPR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BMPR2 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          BMPR2 Antibody in Western Blot (WB)
          BMPR2 Antibody in Western Blot (WB)
          BMPR2 Antibody in Flow Cytometry (Flow)
          BMPR2 Antibody in Immunoprecipitation (IP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          BMPR2 Polyclonal Antibody

          Product Details

          19087-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000-1:6,000
          View 4 publications 4 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunocytochemistry (ICC/IF)

          -
          View 1 publication 1 publication

          Flow Cytometry (Flow)

          0.2 µg/1x10^6 cells
          -

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          BMPR2 Fusion Protein Ag5868 (172-504 aa encoded by BC052985)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.37 mg/mL

          Amount

          55.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          This antibody recognizes endogenous levels of total BMPR2 protein, including 115 kDa precursor, 75 kDa mature form and 150 kDa dimer.

          Immunogen sequence: YRMLTGDRK QGLHSMNMME AAASEPSLDL DNLKLLELIG RGRYGAVYKG SLDERPVAVK VFSFANRQNF INEKNIYRVP LMEHDNIARF IVGDERVTAD GRMEYLLVME YYPNGSLCKY LSLHTSDWVS SCRLAHSVTR GLAYLHTELP RGDHYKPAIS HRDLNSRNVL VKNDGTCVIS DFGLSMRLTG NRLVRPGEED NAAISEVGTI RYMAPEVLEG AVNLRDCESA LKQVDMYALG LIYWEIFMRC TDLFPGESVP EYQMAFQTEV GNHPTFEDMQ VLVSREKQRP KFPEAWKENS LAVRSLKETI EDCWDQDAEA RLTAQCAEER MAEL (172-504 aa encoded by B C052985)

          Target Information

          BMPR2 is a member of the bone morphogenetic protein (BMP) receptor family of transmembrane serine/threonine kinases. The ligands of this receptor are BMPs that are involved in endochondral bone formation and embryogenesis. The loss of interaction and lack of phosphorylation of TCTEL1 by BMPR2 may contribute to the pathogenesis of primary pulmonary hypertension (PPH) and BMPR2 also plays an essential role in human T-cell differentiation. BMPs are involved in endochondral bone formation and embryogenesis. These proteins transduce their signals through the formation of heteromeric complexes of two different types of serine (threonine) kinase receptors: type I receptors of about 50-55 kDa and type II receptors of about 70-80 kDa. Type II receptors bind ligands in the absence of type I receptors, but they require their respective type I receptors for signaling, whereas type I receptors require their respective type II receptors for ligand binding. Mutations in BMPR2 have been associated with primary pulmonary hypertension, both familial and fenfluramine-associated, and with pulmonary venoocclusive disease.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: BMP type II receptor; BMP type-2 receptor; BMP-2/4 type II receptor; bone morphogenetic protein receptor type II; bone morphogenetic protein receptor, type II (serine/threonine kinase); bone morphogenic protein receptor, type II (serine/threonine kinase); Ser/Thr kinase receptor; type II activin receptor-like kinase; type II receptor for bone morphogenetic protein-4; unnamed protein product

          View more View less

          Gene Aliases: 2610024H22Rik; BMP-2; BMPR-2; BMPR-II; BMPR3; BMPRII; BMR2; BRK-3; Gm20272; POVD1; PPH1; T-ALK

          View more View less

          UniProt ID: (Human) Q13873, (Mouse) O35607

          View more View less

          Entrez Gene ID: (Human) 659, (Mouse) 12168, (Rat) 140590

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity transmembrane receptor protein serine/threonine kinase activity transforming growth factor beta-activated receptor activity protein binding ATP binding activin receptor activity, type II BMP binding cadherin binding BMP receptor activity protein tyrosine kinase binding growth factor binding nucleotide binding
          Process(es)
          angiogenesis regulation of cell growth blood vessel development osteoblast differentiation mesoderm formation cell fate specification maternal placenta development endothelial cell proliferation vasculature development lymphangiogenesis blood vessel remodeling chondrocyte development negative regulation of systemic arterial blood pressure outflow tract septum morphogenesis outflow tract morphogenesis aortic valve development pulmonary valve development atrioventricular valve morphogenesis mitral valve morphogenesis tricuspid valve morphogenesis endocardial cushion development transmembrane receptor protein serine/threonine kinase signaling pathway negative regulation of cell proliferation cellular response to starvation tissue development anterior/posterior pattern specification positive regulation of gene expression positive regulation of epithelial cell migration regulation of lung blood pressure cell differentiation proteoglycan biosynthetic process negative regulation of cell growth lung development positive regulation of bone mineralization BMP signaling pathway activin receptor signaling pathway positive regulation of locomotion regulation of cell proliferation positive regulation of cell differentiation positive regulation of osteoblast differentiation positive regulation of ossification negative regulation of vasoconstriction positive regulation of transcription from RNA polymerase II promoter lung alveolus development negative regulation of smooth muscle cell proliferation cardiac muscle tissue development positive regulation of axon extension involved in axon guidance stem cell differentiation limb development endochondral bone morphogenesis positive regulation of SMAD protein import into nucleus ventricular septum morphogenesis atrial septum morphogenesis lung vasculature development lymphatic endothelial cell differentiation artery development venous blood vessel development positive regulation of cartilage development retina vasculature development in camera-type eye pharyngeal arch artery morphogenesis cellular response to growth factor stimulus cellular response to BMP stimulus endothelial cell apoptotic process negative regulation of chondrocyte proliferation semi-lunar valve development regulation of multicellular organismal development negative regulation of cell proliferation involved in heart morphogenesis retina development in camera-type eye
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-x9r6b:80/100.66.78.146:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0