    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
  • Services
    • 360° CDMO and CRO Solutions
          • Primary Antibodies ›
          • HA Tag Antibodies
          Bioss
          HA-Tag Monoclonal Antibody (3C8), Biotin
          View all (65) HA Tag antibodies
          Tech Support
          Cite HA-Tag Monoclonal Antibody (3C8), Biotin
          Additional Information:
          {{banner.description}}
          Product Details
          BSM-33003M-BIOTIN
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:300-1:5,000
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG
          Class
          Monoclonal
          Type
          Antibody
          Clone
          3C8
          Immunogen
          KLH conjugated HA Tag peptide
          Conjugate
          Biotin Biotin Biotin
          • Unconjugated
          • HRP
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          TBS with 1% BSA, 50% glycerol
          Contains
          0.03% ProClin 300
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
          Bioinformatics
          Protein Aliases: HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin
          Hong Kong flag icon
          Hong Kong
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-pgj68:80/100.66.79.163:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline