    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
          • Primary Antibodies ›
          • Cytokeratin 18 Antibodies
          Bioss
          CK18 Monoclonal Antibody (10H2), Cyanine5.5
          View all (149) Cytokeratin 18 antibodies
          Tech Support
          Cite CK18 Monoclonal Antibody (10H2), Cyanine5.5
          Additional Information:
          {{banner.description}}
          CK18 Monoclonal Antibody (10H2), Cyanine5.5
          Product Details
          BSM-33103M-CY55
          Applications
          Tested Dilution
          Publications
          Immunocytochemistry (ICC/IF)
          1:50-1:200
          -
          Product Specifications
          Species Reactivity
          Human, Mouse
          Host/Isotype
          Mouse / IgG
          Class
          Monoclonal
          Type
          Antibody
          Clone
          10H2
          Immunogen
          KLH conjugated synthetic peptide derived from human CK18
          3D Epitope / Immunogen
          Conjugate
          Cyanine5.5 Cyanine5.5 Cyanine5.5
          • Unconjugated
          • Alexa Fluor 488 
          • Alexa Fluor 555 
          • Alexa Fluor 594 
          • Alexa Fluor 647 
          • Alexa Fluor 680 
          • Alexa Fluor 750 
          • Biotin 
          • Cyanine3
          • Cyanine5 
          • Cyanine7
          • FITC
          • HRP
          • Request custom conjugation
          Excitation/Emission Max
          651/670 nm
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein G
          Storage buffer
          TBS with 1% BSA, 50% glycerol
          Contains
          0.03% ProClin 300
          Storage conditions
          -20°C or -80°C if preferred
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          Cytokeratin 18 (Keratin 18, KRT18, type I cytoskeletal 18) belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Further, cytokeratin 18 is found primarily in non-squamous epithelia and is present in a majority of adenocarcinomas and ductal carcinomas, but not in squamous cell carcinomas. Hepatocellular carcinomas express only cytokeratins 8 and 18. Mutations in the cytokeratin 18 gene have been linked to cryptogenic cirrhosis. Two transcript variants encoding the same protein have been found for the cytokeratin 18 gene. Cytokeratin subtype expression patterns are used to an increasing extent in the distinction of different types of epithelial malignancies. The cytokeratin antibodies are not only of assistance in the differential diagnosis of tumors using immunohistochemistry on tissue sections, but are also a useful tool in cytopathology and flow cytometric assays.
          Bioinformatics
          Protein Aliases: Cell proliferation-inducing gene 46 protein; CK 18; CK-18; cytokeratin 18; Cytokeratin endo B; Cytokeratin-18; Cytokeratin18; Endo B; K 18; K18; keratin 18, type I; keratin complex 1, acidic, gene 18; Keratin D; Keratin, type I cytoskeletal 18; Keratin-18; Keratin18; KRT 18; ORF; PIG46; type I cytoskeletal 18; unnamed protein product
          View more View less
          Gene Aliases: CK-18; CK18; CYK18; K18; Kerd; Krt1-18; KRT18; PIG46
          View more View less
          UniProt ID: (Human) P05783, (Mouse) P05784
          View more View less
          Entrez Gene ID: (Human) 3875, (Mouse) 16668
          View more View less
          Function(s)
          RNA binding structural molecule activity protein binding scaffold protein binding cadherin binding involved in cell-cell adhesion poly(A) RNA binding
          Process(es)
          anatomical structure morphogenesis tumor necrosis factor-mediated signaling pathway Golgi to plasma membrane protein transport negative regulation of apoptotic process intermediate filament cytoskeleton organization extrinsic apoptotic signaling pathway hepatocyte apoptotic process cell-cell adhesion Golgi to plasma membrane CFTR protein transport
          • News News
          • Responsibility Responsibility
          Chile flag icon
          Chile
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline