    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
  • Services
    • 360° CDMO and CRO Solutions
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
          • Primary Antibodies ›
          • HA Tag Antibodies
          Bioss
          HA-Tag Monoclonal Antibody (4D1)
          View all (102) HA Tag antibodies
          Tech Support
          Cite HA-Tag Monoclonal Antibody (4D1)
          Additional Information:
          {{banner.description}}
          • Antibody Testing Data (4)
          HA-Tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          HA-Tag Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          FIGURE: 1 / 4
          HA-Tag Antibody (BSM-33157M) in WB
          HA-Tagged Fusion Protein Overexpression E.coli Lysates (Cat#: bs-41230P) probed with HA-Tag (4D1) Monoclonal Antibody, Unconjugated (bsm-33157M) at 1:1000 dilution and 4°C overnight incubation. Followed by conjugated secondary antibody incubation at 1:20000 for 60 min at 37°C. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product
          {{$ctrl.videoDesc}}
          {{$ctrl.videoLongDesc}}
          HA-Tag Antibody in Western Blot (WB)
          HA-Tag Antibody in Western Blot (WB)
          HA-Tag Antibody in Western Blot (WB)
          HA-Tag Antibody in Western Blot (WB)
          Product Details
          BSM-33157M
          Applications
          Tested Dilution
          Publications
          Western Blot (WB)
          1:300-1:5,000
          -
          Product Specifications
          Species Reactivity
          Tag
          Host/Isotype
          Mouse / IgG
          Class
          Monoclonal
          Type
          Antibody
          Clone
          4D1
          Immunogen
          KLH conjugated HA Tag peptide
          Conjugate
          Unconjugated Unconjugated Unconjugated
          • Biotin 
          • HRP
          • Request custom conjugation
          Form
          Liquid
          Concentration
          1 mg/mL
          Amount
          100 µg
          Purification
          Protein A
          Storage buffer
          TBS, pH 7.4, with 50% glycerol, 1% BSA
          Contains
          0.03% ProClin 300
          Storage conditions
          -20°C, Avoid Freeze/Thaw Cycles
          Shipping conditions
          Ambient (domestic); Wet ice (international)
          Target Information
          HA Tag (hemagglutinin tag) is an epitope tag that provides a method to localize gene products in a variety of cell types, study the topology of proteins and protein complexes, identify associated proteins, and characterize newly identified, low abundance or poorly immunogenic proteins when protein specific antibodies are not available. HA (hemagglutinin) is a surface glycoprotein derived from influenza hemagglutinin, and is commonly expressed as a tag in conjunction with proteins to aid in identification and purification of the tagged protein for functional analysis. HA tag antibodies can be used to facilitate protein detection or co-immunopreciptation of tagged proteins.
          Bioinformatics
          Protein Aliases: HA; HA Epitope Tag; HA Tag; HA-tag; hemagglutinin
          Taiwan flag icon
          Taiwan
          Your items have has been added!
          Host server : magellan-search-green-b49b87d85-n9r8p:80/100.66.76.150:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline