Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Bif1 Antibodies

          Proteintech

          Bif-1 Polyclonal Antibody

          4 References
          View all (15) Bif1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Bif-1 Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (16)
          Bif-1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Bif-1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 16

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Bif-1 Antibody (15422-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells, using SH3GLB1 antibody 15422-1-AP at 1:25 dilution and Rhodamine-labeled goat anti-rabbit IGG (red). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Bif-1 Antibody in Immunocytochemistry (ICC/IF)
          Bif-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Bif-1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Antibody in Western Blot (WB)
          Bif-1 Polyclonal Antibody

          Product Details

          15422-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          View 3 publications 3 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:10-1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Bif-1 Fusion Protein Ag7710 (1-365 aa encoded by BC007455)
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.29 mg/mL

          Amount

          43.5 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MNIMDFNVK KLAADAGTFL SRAVQFTEEK LGQAEKTELD AHLENLLSKA ECTKIWTEKI MKQTEVLLQP NPNARIEEFV YEKLDRKAPS RINNPELLGQ YMIDAGTEFG PGTAYGNALI KCGETQKRIG TADRELIQTS ALNFLTPLRN FIEGDYKTIA KERKLLQNKR LDLDAAKTRL KKAKAAETRN SSEQELRITQ SEFDRQAEIT RLLLEGISST HAHHLRCLND FVEAQMTYYA QCYQYMLDLQ KQLGSFPSNY LSNNNQTSVT PVPSVLPNAI GSSAMASTSG LVITSPSNLS DLKECSGSRK ARVLYDYDAA NSTELSLLAD EVITVFSVVG MDSDWLMGER GNQKGKVPIT YLELLN (1-365 aa encoded by B C007455)

          Target Information

          Bif1 gene encodes a protein similar to a protein in rodents which is induced by bone morphogenic protein 2 in vitro and may be involved in BMP2-induced transcription.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Bax-binding protein; Bax-interacting factor 1; Bif-1; BIF1; endophilin b1; Endophilin-B1; KIAA0491; protein phosphatase 1, regulatory subunit 70; SH3 domain-containing GRB2-like protein B1; SH3-containing protein SH3GLB1; SH3-domain GRB2 like endophilin B1; SH3-domain GRB2-like B1 (endophilin); sh3glb1 {ECO:0000312|EMBL:AAH79085.1}; testicular tissue protein Li 172; unnamed protein product

          View more View less

          Gene Aliases: AA409932; AI314629; AU015566; Bif-1; CGI-61; dJ612B15.2; KIAA0491; mKIAA0491; PPP1R70; SH3GLB1

          View more View less

          UniProt ID: (Human) Q9Y371, (Rat) Q6AYE2, (Mouse) Q9JK48

          View more View less

          Entrez Gene ID: (Human) 51100, (Rat) 292156, (Mouse) 54673

          View more View less

          Function(s)
          fatty acid binding protein binding lipid binding lysophosphatidic acid acyltransferase activity identical protein binding protein homodimerization activity cadherin binding phosphatidylinositol 3-kinase activator activity
          Process(es)
          phosphatidic acid biosynthetic process autophagy apoptotic process phospholipid biosynthetic process positive regulation of autophagy regulation of macroautophagy positive regulation of protein complex assembly regulation of protein stability regulation of cytokinesis receptor catabolic process cellular response to amino acid starvation cellular response to glucose starvation autophagic cell death 'de novo' posttranslational protein folding membrane organization membrane fission positive regulation of membrane tubulation positive regulation of establishment of protein localization to mitochondrion protein localization to vacuolar membrane positive regulation of protein targeting to mitochondrion positive regulation of autophagosome assembly positive regulation of protein oligomerization protein oligomerization positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway mitochondrion organization positive regulation of phosphatidylinositol 3-kinase activity negative regulation of intrinsic apoptotic signaling pathway by p53 class mediator positive regulation of intrinsic apoptotic signaling pathway by p53 class mediator
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Chile flag icon
          Chile

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-9fm2r:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline