Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Biglycan Antibodies

          Proteintech

          Biglycan Polyclonal Antibody

          25 References
          View all (18) Biglycan antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Biglycan Polyclonal Antibody

          Additional Information:
          {{banner.description}}
          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (22)
          Biglycan Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Biglycan Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 22

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Biglycan Antibody (16409-1-AP) in ICC/IF

          Immunofluorescent analysis of HepG2 cells using 16409-1-AP (Biglycan antibody) at dilution of 1:50 and Alexa Fluor 488-conjugated AffiniPure Goat Anti-Rabbit IGG (H+L). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Biglycan Antibody in Immunocytochemistry (ICC/IF)
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Biglycan Antibody in Western Blot (WB)
          Biglycan Antibody in Western Blot (WB)
          Biglycan Antibody in Western Blot (WB)
          Biglycan Antibody in Western Blot (WB)
          Biglycan Antibody in Western Blot (WB)
          Biglycan Antibody in Western Blot (WB)
          Biglycan Antibody in Immunoprecipitation (IP)
          Biglycan Polyclonal Antibody

          Product Details

          16409-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          View 12 publications 12 publications

          Immunohistochemistry (IHC)

          -
          View 7 publications 7 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          -

          Immunocytochemistry (ICC/IF)

          1:20-1:200
          View 5 publications 5 publications

          Immunoprecipitation (IP)

          0.5-4.0 µg
          -

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Pig, Rat

          Published species

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Biglycan Fusion Protein Ag0683 (1-368 aa encoded by BC004244)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.22 mg/mL

          Amount

          33 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MWPLWRLVS LLALSQALPF EQRGFWDFTL DDGPFMMNDE EASGADTSGV LDPDSVTPTY SAMCPFGCHC HLRVVQCSDL GLKSVPKEIS PDTTLLDLQN NDISELRKDD FKGLQHLYAL VLVNNKISKI HEKAFSPLRK LQKLYISKNH LVEIPPNLPS SLVELRIHDN RIRKVPKGVF SGLRNMNCIE MGGNPLENSG FEPGAFDGLK LNYLRISEAK LTGIPKDLPE TLNELHLDHN KIQAIELEDL LRYSKLYRLG LGHNQIRMIE NGSLSFLPTL RELHLDNNKL ARVPSGLPDL KLLQVVYLHS NNITKVGVND FCPMGFGVKR AYYNGISLFN NPVPYWEVQP ATFRCVTDRL AIQFGNYKK (1-368 aa encoded by BC004244 )

          Target Information

          BGN is a small cellular or pericellular matrix proteoglycan that is closely related in structure to two other small proteoglycans, decorin and fibromodulin. The protein and decorin are thought to be the result of a gene duplication. Decorin contains one attached glycosaminoglycan chain, while this protein probably contains two chains. For this reason, this protein is called biglycan. This protein is thought to function in connective tissue metabolism by binding to collagen fibrils and transferring growth factor-beta. It may promote neuronal survival.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Biglycan; bone/cartilage proteclycan 1; Bone/cartilage proteoglycan I; bone/cartilage proteoglycan-I; dermatan sulphate proteoglycan I; PG S1; PG-S1; proteoglycan I precursor; small leucine-rich protein 1A; Small proteoglycan I (biglycan), bone (BSPG1) (bone/cartilage proteclycan 1 precursor); unnamed protein product

          View more View less

          Gene Aliases: BG; BGN; BSPG1; DSPG1; MRLS; PG-S1; PGI; SEMDX; SLRR1A

          View more View less

          UniProt ID: (Human) P21810, (Mouse) P28653, (Rat) P47853

          View more View less

          Entrez Gene ID: (Pig) 397396, (Human) 633, (Mouse) 12111, (Rat) 25181

          View more View less

          Function(s)
          extracellular matrix structural constituent conferring compression resistance extracellular matrix structural constituent protein binding glycosaminoglycan binding cytokine binding extracellular matrix binding
          Process(es)
          peptide cross-linking via chondroitin 4-sulfate glycosaminoglycan blood vessel remodeling bone development articular cartilage development
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline