FIGURE: 1 / 4
This antibody is predicted to react with mouse, rat, Xenopus laevis and zebrafish based on sequence homology.
This gene encodes a protein involved in spindle checkpoint function. The encoded protein contains four WD repeat domains and has sequence similarity with the yeast BUB3 protein. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.